Skip to Content
Merck
All Photos(3)

Key Documents

F3754

Sigma-Aldrich

Ammonium iron(II) sulfate hexahydrate

ReagentPlus®, ≥98%

Synonym(s):

Ammonium ferrous sulfate hexahydrate, Ammonium iron(II) sulfate, Mohr’s salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Fe(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
392.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥98%
form:
powder or crystals

Quality Level

product line

ReagentPlus®

Assay

≥98%

form

powder or crystals

pH

3-5 (20 °C, 50 g/L)

mp

100 °C (dec.) (lit.)

SMILES string

N.N.O.O.O.O.O.O.[Fe++].OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Fe.2H3N.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h;2*1H3;2*(H2,1,2,3,4);6*1H2/q+2;;;;;;;;;;/p-2

InChI key

MQLVWQSVRZVNIP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Semwal et al.
Physics in medicine and biology, 47(12), N179-N183 (2002-07-18)
The FBX aqueous chemical dosimeter contains 0.2 mol m(-3) ferrous ammonium sulphate, 5.0 mol m(-3) benzoic acid and 0.20 mol m(-3) xylenol orange in 40.0 mol m(-3) sulphuric acid. The dosimeter can measure photon and electron doses in the range
Cinzia Signorini et al.
Pediatric research, 63(3), 287-291 (2008-02-22)
Nonprotein-bound iron (NPBI) and F2-isoprostanes, reliable markers of oxidative stress, are increased in plasma of newborns and inversely correlated to the gestational age. Because NPBI represents a pro-oxidant stimulus in plasma, we test the hypothesis that the entity of lipid
Mayara Marinovic Baccan et al.
Journal of inorganic biochemistry, 107(1), 34-39 (2011-12-17)
The antioxidant activity of flavonoids may involve their ability to complex body iron in non-redox-active forms. In this study, it was found that the catechol flavonoids rutin and quercetin are able to suppress redox-active labile plasma iron (LPI) in both
Xianwen Mi et al.
Analytical chemistry, 84(2), 939-946 (2011-12-07)
The key factors that control the spread and mortality rate of tuberculosis (TB) are rapid detection and diagnosis. However, the current detection of Mycobacterium tuberculosis (M. tuberculosis) cannot meet the recommended requirements for clinical diagnosis in turnaround time. In this
Hyung Lee et al.
Journal of neurosurgical anesthesiology, 17(3), 144-148 (2005-07-23)
This study compared the effectiveness of propofol with that of trolox and melatonin for reduction of lipid peroxidation in vitro. Lipid peroxidation was induced by addition of kainic acid (KA; 10 mM), hydrogen peroxide (H2O2; 10 mM), or ferrous ammonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service