Skip to Content
Merck
All Photos(3)

Key Documents

336378

Sigma-Aldrich

Lead(II) carbonate

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbCO3
CAS Number:
Molecular Weight:
267.21
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
grade:
ACS reagent
form:
powder

grade

ACS reagent

form

powder

reaction suitability

reagent type: catalyst
core: lead

impurities

≤0.02% Insoluble matter in dilute HOAc

anion traces

NO3- and NO2-: ≤0.005%
chloride (Cl-): ≤0.002%

cation traces

Ca: ≤0.01%
Cd: ≤0.002%
Fe: ≤0.005%
K: ≤0.02%
Na: ≤0.05%
Zn: ≤0.003%

SMILES string

[PbH2++].[O-]C([O-])=O

InChI

1S/CH2O3.Pb/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

MFEVGQHCNVXMER-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yasuo Izumi et al.
Analytical chemistry, 74(15), 3819-3823 (2002-08-15)
The local structure of trace amounts of lead in an adsorbent matrix that contains a high concentration of iron and magnesium (Mg6Fe2(OH)16(CO3) x 3H2O) was successfully monitored by means of X-ray absorption fine structure spectroscopy combined with fluorescence spectrometry. A
Brian Gulson et al.
Environmental research, 112, 100-110 (2011-09-20)
In late 2006, the seaside community in Esperance Western Australia was alerted to thousands of native bird species dying. The source of the lead (Pb) was determined by Pb isotopes to derive from the handling of Pb carbonate concentrate through
Ceruse: a skin lightening cosmetic at the king's court.
Prescrire international, 22(135), 55-55 (2013-02-28)
Xin Xu et al.
Huan jing ke xue= Huanjing kexue, 28(7), 1644-1648 (2007-09-26)
The sequestration of CO2 and carbonation process for the stabilization of fresh fly ash from municipal solid waste (MSW) incinerator were investigated. The purpose of this paper is to evaluate the influence of fundamental parameters affecting the stabilization of heavy
J Nunes et al.
Toxicologic pathology, 29(4), 451-457 (2001-09-19)
To determine if additive or synergistic toxic effects would occur, adult male rats were exposed orally to lead carbonate (2,000 mg/kg) for 9 days before trichloroethylene (TCE), 2,000 mg/kg, was given concurrently for an additional 7 days. Comparisons were made

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service