Skip to Content
Merck
All Photos(3)

Documents

232084

Sigma-Aldrich

Molybdic acid

≥85.0% MoO3 basis, ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent

vapor density

6.2 (vs air)

Assay

≥85.0% MoO3 basis

form

powder

reaction suitability

reagent type: catalyst
core: molybdenum

impurities

≤0.01% insol. dil. NH4OH

solubility

water: insoluble

density

3.1 g/mL at 25 °C (lit.)

anion traces

AsO43-, PO43- and SiO2: ≤0.001%
chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.2%

cation traces

heavy metals: ≤0.003%

SMILES string

O[Mo](O)(=O)=O

InChI

1S/Mo.2H2O.2O/h;2*1H2;;/q+2;;;;/p-2

InChI key

VLAPMBHFAWRUQP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Molybdic acid is the hydrated form of molybdenum trioxide. It is used in the synthesis Froehde reagent.

Application

Molybdic acid can be used:
  • In the preparation of silica-supported molybdic acid catalyst for the synthesis of pyrano [2,3-c] chromene derivatives by condensation of aldehydes, malononitrile, and 4-hydroxycoumarin.
  • In the synthesis of Froehde reagent for the presumptive identification of alkaloids like opioids.
  • As a molybdenum source in the preparation of MoSe2.

Other Notes

Consists largely of an ammonium molybdate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of high-temperature shock-wave compression on few-layer MoS2, WS2 and MoSe2.
Vasu K, et al.
Chemical Physics Letters, 582, 105-109 (2013)
Hybrid Fabric/Molybdic Acid Modified Phenolic Resin Composites with Improved Anti-wear Properties.
Yang M, et al.
Tribology Transactions (2015)
Made by molybdenum.
Lennartson A.
Nature Chemistry, 6(8), 746-746 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service