Skip to Content
Merck
All Photos(1)

Key Documents

QC1364

Supelco

Anions

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-011

concentration

in H2O

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service