Skip to Content
Merck
All Photos(1)

Key Documents

QC1051

Supelco

Complex Nutrients

certified reference material

Synonym(s):

Compound Nutrient Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-028-2

concentration

in H2O

technique(s)

AAS: suitable
ICP: suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application


  • Optimizing cell culture media with complex nutrients: Research highlighted the development of optimized cell culture media using complex nutrients to enhance cellular functions and viability, providing a scalable solution for biotechnological and pharmaceutical applications (Krynická and Komenda, 2024).

  • Complex nutrients in public health education: A study emphasized the importance of complex nutrients in daily diets and their role in supporting immune function, stressing the need for primary healthcare providers to educate the community about nutritional self-care (Calder et al., 2023).

  • Complex nutrients for metabolic regulation in oncology: Investigations into the role of complex nutrients in regulating fatty acid delivery to cancer metastases suggest their potential therapeutic applications in oncological research (Edwards et al., 2024).

  • Marine-derived complex nutrients for livestock health: A study on marine-derived Ca-Mg complex nutrients showed significant improvements in lipid and glucose metabolism, serum metabolites, and stress hormone levels in livestock, indicating their value in animal nutrition and biotechnology (Cho et al., 2023).

  • Complex nutrients in radiation therapy: Research explored the application of complex nutrients in mitigating gastrointestinal acute radiation syndrome, demonstrating their potential as medical countermeasures in radiation exposure scenarios (Winters et al., 2024).

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service