PHR3877
Phytic Acid Dodecasodium Salt Anhydrous
certified reference material, pharmaceutical secondary standard
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
API family
phytic acid solution
packaging
pkg of 500MG
application(s)
pharmaceutical
storage temp.
2-30°C
General description
Analysis Note
Other Notes
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service