Skip to Content
Merck
All Photos(1)

Key Documents

PHR1865

Supelco

Duloxetine Hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-Duloxetine hydrochloride, (γS)-N-Methyl-γ-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride, (+)-(S)-N-Methyl-γ-(1-naphthyloxy)-2-thiophenepropylamine hydrochloride, (+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, LY-248686 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NOS · HCl
CAS Number:
Molecular Weight:
333.88
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001453
traceable to USP 1229817

API family

duloxetine

form

powder

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

Cl.CNCC[C@H](Oc1cccc2ccccc12)c3cccs3

InChI

1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m0./s1

InChI key

BFFSMCNJSOPUAY-LMOVPXPDSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Duloxetine Hydrochloride, a thiophene derivative, belongs to the class of selective serotonin-norepinephrine reuptake inhibitors (SNRI), used for its anti-depressant properties.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Development of two voltammetry methods using carbon paste electrode, for the trace analysis of duloxetine hydrochloride in pharmaceutical dosage forms and human serum samples
  • Separation and estimation of curcumin and duloxetine hydrochloride by reversed-phase high-performance liquid chromatography (RP-HPLC) from bulk and pharmaceutical formulations
  • Determination of duloxetine in pure form by a UV-Vis spectrophotometric method, in accordance with the ICH Q2R1 guidelines
  • Multi-analysis of antidepressant drugs― vilazodone hydrochloride, agomelatine, and duloxetine hydrochloride along with vitamin B12 in their bulk, pharmaceutical formulations, and human urine samples using HPLC

Biochem/physiol Actions

Duloxetine hydrochloride is a dual serotonin/norepinephrine reuptake inhibitor (SNRI), widely used clinically as an antidepressant and anxiolytic.
Duloxetine hydrochloride is a dual serotonin/norepinephrine reuptake inhibitor (SNRI); antidepressant; anxiolytic.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1689 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validated reversed-phase high-performance liquid chromatography method for simultaneous estimation of curcumin and duloxetine hydrochloride in tablet and self-nanoemulsifying drug delivery systems
Kumar B, et al.
Journal of Pharmacy Research, 11, 1166-1166 (2017)
Development and validation of UV-Visible spectrophotometric method for determination of Duloxetine
Samal, LIPSA and Prusty, AMARESH
International Journal of Pharmacy and Pharmaceutical Sciences, 11, 27-31 (2019)
Trace determination of duloxetine HCl in formulation and spiked human serum at a carbon paste electrode
Hassanein AM, et al.
American Journal of Analytical Chemistry, 8, 708-725 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service