Skip to Content
Merck
All Photos(1)

Key Documents

PHR1506

Supelco

Metformin Related Compound C

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

N,N-Dimethyl-1,3,5-triazine-2,4,6-triamine, N,N-Dimethyl-[1,3,5]-triazine-2,4,6-triamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N6
CAS Number:
Molecular Weight:
154.17
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1396342

API family

metformin

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(C)(C)c1nc(nc(n1)N)N

InChI

1S/C5H10N6/c1-11(2)5-9-3(6)8-4(7)10-5/h1-2H3,(H4,6,7,8,9,10)

InChI key

IEFWDQQGFDLKFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

It is an impurity of the widely used anti-diabetic drug, metformin, that belongs to the biguanide class of compounds.

Application

This pharmaceutical secondary standard can also be used as follows:

  • Simultaneous determination of vildagliptin, metformin, and metformin-related compounds (A, B, and C) in tablets using a high-performance liquid chromatography-tandem mass spectrometry method (HPLC-MS/MS)
  • Development of a hydrophilic interaction liquid chromatography method combined with diode array detection (HILIC-DAD) for the simultaneous quantification of impurities in the combined dosage tablet of rosuvastatin and metformin
  • Analysis of metformin and its related impurities in bulk samples by normal phase high-performance liquid chromatographic (HPLC) method

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2896 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new rapid and economic liquid chromatographic method for simultaneous determination of meglitinides with metformin: application in the presence of metformin and repaglinide impurities and related compounds
El-Zaher AA, et al.
Journal of the Iranian Chemical Society, 15, 61-74 (2018)
Development of normal phase high-performance liquid chromatographic method for determination of metformin and its related impurities in bulk material
Reid IOA
The pharma innovation journal, 6, 157-157 (2017)
HPLC-MS/MS method for simultaneous quantification of vildagliptin, metformin, and metformin-related compounds in tablets
Uber CP, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 6, 203-207 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service