Skip to Content
Merck
All Photos(1)

Key Documents

NIST3248

Ginkgo-containing tablets

NIST® SRM® 3248

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in validating analytical methods for the determination of flavonoids, terpene lactones, and toxic elements in ginkgo-containing tablets and similar matrices. This SRM can also be used for quality assurance when assigning values to in-house control materials. SRM 3248 is part of a suite of ginkgo dietary supplement SRMs that have been developed to cover a range of natural matrices and analyte levels. A unit of SRM 3248 consists of five bottles, each containing approximately 1 g of ground tablets. For more information, please refer to the SDS and COA.

SRM 3248_cert

SRM 3248_SDS

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: Bilobalide, Ginkgolide A, Ginkgolide B, Ginkgolide C, Ginkgolide J, Isorhamnetin, Kaempferol, Quercetin, Total Aglycones, Total Terpene Lactones
Trace Elements: Arsenic (As), Cadmium (Cd), Lead (Pb), Mercury (Hg)

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service