Skip to Content
Merck
All Photos(1)

Key Documents

N0399000

Neohesperidin dihydrochalcone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(4-((2-O-[6-Deoxy-α-L-mannopyranosyl]-β-D-glucopyranosyl)oxy)-2,6-dihydroxyphenyl)-3-[3-hydroxy-4-methoxyphenyl]-1-propanone, NHDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36O15
CAS Number:
Molecular Weight:
612.58
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

neohesperidin

manufacturer/tradename

EDQM

mp

156-158 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

COc1ccc(CCC(=O)c2c(O)cc(O[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)cc2O)cc1O

InChI

1S/C28H36O15/c1-11-21(34)23(36)25(38)27(40-11)43-26-24(37)22(35)19(10-29)42-28(26)41-13-8-16(32)20(17(33)9-13)14(30)5-3-12-4-6-18(39-2)15(31)7-12/h4,6-9,11,19,21-29,31-38H,3,5,10H2,1-2H3/t11-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1

InChI key

ITVGXXMINPYUHD-CUVHLRMHSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHL8228975041N8757
grade

pharmaceutical primary standard

grade

primary reference standard

grade

analytical standard

grade

-

format

neat

format

-

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

-

API family

neohesperidin

API family

-

API family

-

API family

-

mp

156-158 °C (lit.)

mp

156-158 °C (lit.)

mp

156-158 °C (lit.)

mp

156-158 °C (lit.)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Neohesperidin dihydrochalcone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Lihua Hu et al.
    Chemico-biological interactions, 213, 51-59 (2014-02-18)
    The purpose of this study was to investigate the possible hepatoprotective effects of neohesperidin dihydrochalcone (NHDC) on carbon tetrachloride (CCl4)-induced acute oxidative injury in vivo and in vitro. In a mouse model, intraperitoneal injection of CCl4 resulted in a significant

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service