BCR165
Latex spheres
BCR®, certified reference material, 2 μm particle size
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Agency
BCR®
description
parameter of the calibration line 2.223 0.013
form
particles
manufacturer/tradename
JRC
particle size
2 μm
application(s)
general analytical
format
neat
Looking for similar products? Visit Product Comparison Guide
General description
Nominal 2 μm latex (0.02% solids)
Packaging
Vial containing 2 mL of an aqueous suspension of latex spheres.
Analysis Note
For more information please see:
BCR165
BCR165
Legal Information
BCR is a registered trademark of European Commission
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service