93689
L-Carnitine-(trimethyl-d9) inner salt
analytical standard
Synonym(s):
(R)-3-Carboxy-2-hydroxy-N,N,N-tri(methyl-d3)-1-propanaminium inner salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7D9H6NO3
CAS Number:
Molecular Weight:
170.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
Assay
≥97.0% (HPLC)
optical activity
[α]/D -27.5±3.0°, c = 0.1 in H2O
shelf life
limited shelf life, expiry date on the label
extent of labeling
≥99.5%
application(s)
clinical testing
format
neat
storage temp.
2-8°C
SMILES string
[N+](C[C@H](O)CC(=O)[O-])(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C7H15NO3/c1-8(2,3)5-6(9)4-7(10)11/h6,9H,4-5H2,1-3H3/t6-/m1/s1/i1D3,2D3,3D3
InChI key
PHIQHXFUZVPYII-SAQLGWHKSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service