908614
Grubbs Catalyst® M103
Synonym(s):
Dichloro(3-methyl-2-butenylidene)bis(tricyclohexylphosphine)ruthenium(II), Grubbs Catalyst® M1b (C801)
About This Item
Recommended Products
form
powder
storage temp.
2-8°C
SMILES string
[Ru+2]=CC=C(C)C.P(C6CCCCC6)(C5CCCCC5)C4CCCCC4.P(C3CCCCC3)(C2CCCCC2)C1CCCCC1.[Cl-].[Cl-]
InChI
1S/2C18H33P.C5H8.2ClH.Ru/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-4-5(2)3;;;/h2*16-18H,1-15H2;1,4H,2-3H3;2*1H;/q;;;;;+2/p-2
InChI key
INCDKKABOVOFIV-UHFFFAOYSA-L
Application
Learn more about our metathesis catalysts
Storage and Stability
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service