Skip to Content
Merck
All Photos(1)

Documents

55097

Supelco

Glycine hydrochloride solution

100 mM amino acid in 0.1 M HCl, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5NO2 · HCl
CAS Number:
Molecular Weight:
111.53
Beilstein:
3909617
MDL number:
UNSPSC Code:
85151701
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5%

form

liquid

analyte chemical class(es)

amino acids, peptides, proteins

concentration

100 mM amino acid in 0.1 M HCl

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

application(s)

food and beverages

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.NCC(O)=O

InChI

1S/C2H5NO2.ClH/c3-1-2(4)5;/h1,3H2,(H,4,5);1H

InChI key

IVLXQGJVBGMLRR-UHFFFAOYSA-N

Application

Glycine hydrochloride in combination with 1-allyl-3-methylimidazolium chloride can find applications as an ionic liquid solvent, which can be used to dissolve chitosan and also fabricate chitosan/cellulose blend films with definite proportions of chitosan. It can also be used to treat the condition of achlorhydria.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial blend films of cellulose and chitosan prepared from binary ionic liquid system
Fu R, et al.
Fibers and Polymers, 18, 852-858 (2017)
Jr, M.J.J.
Encyclopedia of Chemical Processing and Design (1977)
Emilie Macé et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 23(1), 7-16 (2014-08-07)
Most inherited retinal dystrophies display progressive photoreceptor cell degeneration leading to severe visual impairment. Optogenetic reactivation of retinal neurons mediated by adeno-associated virus (AAV) gene therapy has the potential to restore vision regardless of patient-specific mutations. The challenge for clinical
Kasper B Hansen et al.
Molecular pharmacology, 84(1), 114-127 (2013-04-30)
N-methyl-d-aspartate (NMDA) receptors are ligand-gated ion channels assembled from GluN1 and GluN2 subunits. We used a series of N-hydroxypyrazole-5-glycine (NHP5G) partial agonists at the GluN2 glutamate binding site as tools to study activation of GluN1/GluN2A and GluN1/GluN2D NMDA receptor subtypes.
Robert J Harvey et al.
Nature reviews. Drug discovery, 12(11), 866-885 (2013-11-01)
Glycine transporters are endogenous regulators of the dual functions of glycine, which acts as a classical inhibitory neurotransmitter at glycinergic synapses and as a modulator of neuronal excitation mediated by NMDA (N-methyl-D-aspartate) receptors at glutamatergic synapses. The two major subtypes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service