Skip to Content
Merck
All Photos(2)

Documents

34213

Supelco

Pazufloxacin

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15FN2O4
CAS Number:
Molecular Weight:
318.30
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

C[C@H]1COc2c3N1C=C(C(O)=O)C(=O)c3cc(F)c2C4(N)CC4

InChI

1S/C16H15FN2O4/c1-7-6-23-14-11(16(18)2-3-16)10(17)4-8-12(14)19(7)5-9(13(8)20)15(21)22/h4-5,7H,2-3,6,18H2,1H3,(H,21,22)/t7-/m0/s1

InChI key

XAGMUUZPGZWTRP-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone
Pazufloxacin is a fluoroquinolone antibiotic, which shows broad spectrum activity in anti-infective chemotherapy.

Application

Pazufloxacin may be used as a reference standard in the determination of pazufloxacin in serum and plasma samples using high performance liquid chromatography coupled with fluorescence detector (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

First liquid chromatography method for the simultaneous determination of levofloxacin, pazufloxacin, gatifloxacin, moxifloxacin and trovafloxacin in human plasma.
Sousa J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 930, 104-111 (2013)
Simultaneous measurement of pazufloxacin, ciprofloxacin, and levofloxacin in human serum by high-performance liquid chromatography with fluorescence detection.
Watabe S, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 878(19), 1555-1561 (2010)
A P Johnson
Current opinion in investigational drugs (London, England : 2000), 1(1), 52-57 (2001-03-16)
Toyama Chemical Co Ltd is developing pazufloxacin (T-3761), an orally active synthetic quinolone antibiotic, which is awaiting registration following successful clinical trials that demonstrated its pharmacological similarities to tosufloxacin (Toyama Chemical Co) with respect to clinical efficacy, adverse effects and
Shinzaburo Minami et al.
Nihon yakurigaku zasshi. Folia pharmacologica Japonica, 122(2), 161-178 (2003-08-02)
Pazufloxacin mesilate (PZFX: Pasil INJECTION, Pazucross INJECTION) is a novel injectable quinolone antibiotic that was discovered by Toyama Chemical Co., Ltd. and codeveloped by Toyama Chemical Co. Ltd. and Mitsubishi Pharma Corporation. Laboratory studies indicated that PZFX reached high plasma
Naoyuki Miyashita et al.
Journal of infection and chemotherapy : official journal of the Japan Society of Chemotherapy, 26(5), 411-417 (2020-02-23)
Legionella species are consistently identified as some of the most common causative agents of severe community-acquired pneumonia (CAP) or nosocomial pneumonia. Although the number of reported Legionella infection cases is gradually increasing in Japan, most cases are diagnosed by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service