Skip to Content
Merck
All Photos(2)

Key Documents

34148

Supelco

Metolachlor OA

PESTANAL®, analytical standard

Synonym(s):

BSTFA, TMCS, N-(2-Ethyl-6-methylphenyl)-N-(2-methoxy-1-methylethyl)oxalamic acid, [(2-Ethyl-6-methylphenyl)(2-methoxy-1-methylethyl)amino]oxo-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCc1cccc(C)c1N(C(C)COC)C(=O)C(O)=O

InChI

1S/C15H21NO4/c1-5-12-8-6-7-10(2)13(12)16(11(3)9-20-4)14(17)15(18)19/h6-8,11H,5,9H2,1-4H3,(H,18,19)

InChI key

LNOOSYCKMKZOJB-UHFFFAOYSA-N

Related Categories

General description

Metolachlor is used for the control of broadleaf weeds and annual grasses primarily in corn, soybean, and sorghum. Active compounds of metolachlor pesticide may be converted by the target insects or plants or may be degraded in the environment to form the corresponding oxanillic (OA) acids. Metolachlor OA is one of the metabolite of metolachlor.

Application

Metolachlor OA may be used as an analytical reference standard for the determination of the analyte in water by solid phase extraction and liquid chromatography/electrospray ionization tandem mass spectroscopy (LC/ESI MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Reagent for trimethylsilyl (TMS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Josef Velisek et al.
The Science of the total environment, 643, 1456-1463 (2018-09-08)
The metolachlor OA is a metabolite of herbicide metolachlor and s-metolachlor. The objective of the present study was to assess the effect metolachlor OA on early life stages of marbled crayfish (Procambarus virginalis). The early life stages of marbled crayfish
Alzbeta Stara et al.
Chemosphere, 224, 616-625 (2019-03-09)
Increasing production of energy crops in Europe, mainly maize and rapeseed, has altered patterns of pesticide use in recent decades. The long-term effects of S-metolachlor (S-M) and of its metabolite metolachlor OA (M-OA) at the environmentally relevant concentration of 4.2 μg L-1
Hidenori Matsukami et al.
The Science of the total environment, 493, 672-681 (2014-07-06)
Chlorinated and non-halogenated organophosphorus flame retardants (OPFRs) including tris(2-chloroisopropyl) phosphate (TCIPP), diethylene glycol bis(di(2-chloroisopropyl) phosphate) (DEG-BDCIPP), triphenyl phosphate (TPHP), and bisphenol A bis(diphenyl phosphate) (BPA-BDPP) have been used increasingly as alternatives to polybrominated diphenyl ethers and other brominated flame retardants.
Robert A Yokley et al.
Analytical chemistry, 74(15), 3754-3759 (2002-08-15)
A Good Laboratory Practices (GLP) validated, multiresidue analytical method is presented for the determination of the chloroacetanilide herbicides metolachlor, acetochlor, and alachlor, the chloroacetamide herbicide dimethenamid, and their respective ethanesulfonic (ESA) and oxanillic (OA) acid degradates in ground and surface
Laura Degani et al.
Analytical and bioanalytical chemistry, 407(6), 1695-1704 (2015-01-19)
The possibility of successfully applying a common GC-MS procedure for identification in one step of all types of dyes from plants of unknown origin and from historical objects is particularly attractive due to the high separation efficiency of the capillary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service