26202
Hamilton® GASTIGHT® syringe, 1700 series, PTFE luer lock
1705TLL, volume 50 μL, PTFE Luer lock, needle size (not included)
All Photos(2)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Recommended Products
material
PTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80920
needle L
, n/a
volume
50 μL
fitting
luer lock (without slots)
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.1 The tip is threaded in a luer lock syringe allowing the needle to be tightly screwed to the tip.
Application
It has been used during measuring phosphine concentration by gas chromatographic-flame photometric detector from the gas collected in fumigation chamber.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Pharmacology for the Prehospital Professional, 54-54 (2009)
Phosphine resistance in eggs of Tribolium castaneum and Plodia interpunctella from almond storage facilities in the Central Valley of California
Gautam, Sandipa G., and George P. Opit.
IOBC-WPRS Bulletin 111, 111, 41-49 (2015)
Don P. Wolf
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Jeffrey S. Guy
Pharmacology for the Prehospital Professional, 54-54 (2009)