Skip to Content
Merck
All Photos(3)

Key Documents

252859

Sigma-Aldrich

Tris Base

99.8-100.1%,≥99.8%, crystalline, ACS reagent

Synonym(s):

Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Product Name

Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%

grade

ACS reagent

Quality Level

Assay

≥99.8%
99.8-100.1%

form

crystalline

technique(s)

affinity chromatography: suitable

impurities

≤0.005% insolubles
≤2% water

color

white

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

cation traces

Fe: ≤5 ppm
heavy metals: ≤5 ppm (ICP)

absorption

passes test

application(s)

clinical research
diagnostic assay manufacturing
microbiology

foreign activity

DNase, none detected
NICKase, none detected
RNase, none detected
protease, none detected

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(hydroxymethyl)aminomethane, commonly known as Trometamol, Tris base, or Trizma base, plays a crucial role in diverse research applications as a biological buffer. It is extensively used in the formulation of buffers like Tris-acetate-EDTA (TAE) and Tris-borate-EDTA (TBE), owing to its optimal pKa of 8.1, making it well-suited for maintaining pH within the physiological range (pH 7 -9) applicable to most living organisms. However, caution might be necessary when studying some proteins, as Tris has the potential to interfere with the activity of certain enzymes. Tris base can be employed both independently as a buffer and as a component in mixed buffer formulations, including Tris-EDTA (TE) buffer, TAE buffer, and TBE buffer, among others. Importantly, Tris base is characterized by its purity, essential stability, and relative non-hygroscopic nature.

In laboratory settings, Tris base is indispensable for the preparation of buffers compatible with biological fluids, acting as a standard pH solution. It facilitates various procedures such as lactate dehydrogenase assays, in situ hybridization, and protein extraction from cells. Its versatility extends to applications in cell culture, biochemistry, and molecular biology labs, where it contributes to studies involving cell membrane permeability and buffer preparation.

Application

Tris(hydroxymethyl)aminomethane has been used:
  • as a buffer solution for lactate dehydrogenase assay, in situ hybridization procedure and protein extraction from cells.[1][2][3]
  • as a buffer in flow cell sample preparation for Raman spectroscopy[4]
  • as a wash buffer in Western blotting[5]
  • as a buffer component in decellularization of tissues[6]

Features and Benefits

  • Efficient buffering within the pH range of 7-9 with a pKa of 8.1 (25 °C)
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Can be used in Cell Biology, and Biochemical research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An optimized lactate dehydrogenase release assay for screening of drug candidates in neuroscience.
Kaja S et al.
Journal of Pharmacological and Toxicological Methods, 73, 1-1 (2015)
Tris-tris-HCl: a standard buffer for use in the physiologic pH range.
R A Durst et al.
Clinical chemistry, 18(3), 206-208 (1972-03-01)
Fernando E Díaz-Manzano et al.
Frontiers in plant science, 7, 966-966 (2016-07-28)
Galls induced by Meloidogyne spp. in plant roots are a complex organ formed by heterogeneous tissues; within them there are 5-8 giant cells (GCs) that root-knot nematodes use for their own nurturing. Subtle regulatory mechanisms likely mediate the massive gene
Jianbin Zhang et al.
Autophagy, 10(5), 901-912 (2014-03-29)
Autophagy is a catabolic process during which cellular components including protein aggregates and organelles are degraded via a lysosome-dependent process to sustain metabolic homeostasis during nutrient or energy deprivation. Measuring the rate of proteolysis of long-lived proteins is a classical
Sylvain Clède et al.
Chemical communications (Cambridge, England), 48(62), 7729-7731 (2012-06-29)
A rhenium tris-carbonyl derivative has been designed to couple infrared and luminescent detection in cells. Both spectroscopies are consistent with one another; they point out the reliability of the present SCoMPI (for Single Core Multimodal Probe for Imaging) for bimodal

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service