Skip to Content
Merck
All Photos(1)

Key Documents

12084

Supelco

Capsaicin

analytical standard

Synonym(s):

8-Methyl-N-vanillyl-trans-6-nonenamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH=CH(CH2)4CONHCH2C6H3-4-(OH)-3-(OCH3)
CAS Number:
Molecular Weight:
305.41
Beilstein:
2816484
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

description

analytical standard for food analysis

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

62-65 °C (lit.)

solubility

H2O: insoluble

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(CNC(=O)CCCC\C=C\C(C)C)ccc1O

InChI

1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/b8-6+

InChI key

YKPUWZUDDOIDPM-SOFGYWHQSA-N

Gene Information

human ... TRPV1(7442)

Looking for similar products? Visit Product Comparison Guide

Application

This analytical standard was used as follows:
  • Determination of capsaicin in samples of chili pepper, black pepper powder, and chili pepper sauce by cyclic voltammetry using an office paper-based carbon black-screen printed electrode (CB-SPE)
  • Development of an electrochemical sensor based on Ag/Ag2O-loaded poly(sodium-4-styrenesulfonate) (PSS) functionalized-reduced graphene oxide (rGO) composite (Ag/Ag2O-PSS-Rgo) to determine capsaicin in chili pepper samples
  • Multi-residue analysis of six capsaicinoids in green bell pepper samples using an online supercritical fluid extraction combined with reverse phase-liquid chromatography-tandem mass spectrometry (RP-LC-MS/MS)
  • Evaluation of an immunoaffinity chromatography (IAC) based cleanup using polyclonal antibodies (pAb) for the determination of capsaicin and dihydrocapsaicin in different types of vegetable oil samples by LC-MS/MS
  • Thin layer chromatographic (TLC) separation and determination of capsaicin in ethanolic extracts of Capsicum annum Linn

Biochem/physiol Actions

Prototype vanilloid receptor agonist. Neurotoxin; activates sensory neurons that give rise to unmyelinated C-fibers, many of which contain substance P. Topical application desensitizes the sensory nerve endings giving a paradoxical antinociceptive effect; systemic administration can be neurotoxic to capsaicin-sensitive cells, especially in newborn animals. Active component of chili peppers.
Prototype vanilloid receptor agonist; neurotoxin. Active component of chili peppers.

Features and Benefits

Superior quality of pure E-Capsaicin, with a guaranteed assay of =99.0% by HPLC.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of capsaicin content in various parts of pepper fruit by liquid chromatography with electrochemical detection.
Supalkova, Veronika, et al.
Acta Chimica Slovenica, 54.1, 55-55 (2007)
S W Hwang et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(11), 6155-6160 (2000-05-24)
Capsaicin, a pungent ingredient of hot peppers, causes excitation of small sensory neurons, and thereby produces severe pain. A nonselective cation channel activated by capsaicin has been identified in sensory neurons and a cDNA encoding the channel has been cloned
Helen Sumner et al.
American journal of respiratory and critical care medicine, 187(9), 943-949 (2013-03-09)
Cough is one of the principal symptoms of chronic obstructive pulmonary disease (COPD) but the potential drivers of cough are likely to be multifactorial and poorly understood. To quantify cough frequency in an unselected group of subjects with COPD and
Emma C Y Hilton et al.
The Journal of allergy and clinical immunology, 132(4), 847-855 (2013-06-20)
Inhaled capsaicin elicits cough reproducibly in human subjects and is widely used in the study of cough and antitussive therapies. However, the traditional end points C2 and C5 (the concentrations of capsaicin inducing at least 2 or 5 coughs, respectively)
Sheena Derry et al.
The Cochrane database of systematic reviews, 2(2), CD007393-CD007393 (2013-03-02)
Topical creams with capsaicin are used to treat peripheral neuropathic pain. Following application to the skin capsaicin causes enhanced sensitivity, followed by a period with reduced sensitivity and, after repeated applications, persistent desensitisation. High-concentration (8%) capsaicin patches were developed to

Questions

  1. How much pungency for standard how to make standard solution

    1 answer
    1. Capsaicin analytical standard is highly soluble in many organic solvents, including DMSO, isopropanol, ethanol, and diethyl ether. The choice of solvent will depend on the analytical technique being used and the sample matrix. Please contact technical service if you need further assistance with your application.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service