Skip to Content
Merck
All Photos(1)

Key Documents

02349

Millipore

HybriScanD Total Bacterial Count

suitable for microbiology

Synonym(s):

FastScan Total Bacterial Count, Total Bacterial Count HybriScanD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85
Pricing and availability is not currently available.

technique(s)

microbe id | in situ hybriziation: suitable

Quality Level

application(s)

agriculture
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

bacteria

General description

HybriScan D kits detect and simultaneously identify microorganisms using rRNA as the detection target. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix, for example food, beverages and water.

Features and Benefits

Sensitivity (cfu/assay): 5000

Legal Information

HybriScan is a trademark of ScanBec GmbH

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Fast molecular screening of beverage, water, wastewater and food for microorganisms

Fast molecular screening of beverage, water, wastewater and food for microorganisms

Fast molecular screening of beverage, water, wastewater and food for microorganisms

Fast molecular screening of beverage, water, wastewater and food for microorganisms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service