Skip to Content
Merck
All Photos(1)

Key Documents

01869

Millipore

Hippurate Strips Kit

suitable for microbiology

Synonym(s):

Hippurate Strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.
Gardnerella vaginalis
Streptococcus spp.

Looking for similar products? Visit Product Comparison Guide

Application

Test for identification of hippurate hydrolase activity of group B streptococci, Campylobacter jejuni, Gardnerella vaginalis and other microorganisms.
The test is based on hydrolysis of substrate (sodium hippurate) by the bacterial enzyme hippurate hydrolase and production of benzoic acid and glycine. Glycine produced by this enzymatic reaction is detected after 24 hours incubation in reaction with chromogen (ninhydrine) and the blue-purple substance is produced.

Components

The kit contains 50 Hippurate Strips (Cat. No. 92472) saturated with sodium hippurate and 350 mg Chromogen in 15 ml Diluent (Cat. No. 01875)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

For microbiologists the most fundamental stain was developed in 1884 by the Danish bacteriologist Hans Christian Gram.

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service