11718096001
Roche
DAB Substrate
dark brown, /black precipitate; Visually evaluated
Synonym(s):
3,3′-diaminobenzidine tetrahydrochloride, DAB
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352204
Recommended Products
manufacturer/tradename
Roche
Quality Level
color
dark brown, /black precipitate; Visually evaluated
storage temp.
−20°C
General description
The metal enhanced DAB Substrate utilizes cobalt chloride and nickel chloride in a special formulation to produce a dark brown/black precipitate in the presence of horseradish peroxidase (HRP).
3, 3′ diaminobenzidine tetrahydrochloride (DAB) is a carcinogen that should be used safely. It is one of the major sensitive substrates for HRP (horseradish peroxidase). DAB is brown or amber in color and becomes insoluble and precipitates over the antigen-antibody site due to the presence of hydrogen peroxide and peroxidase.
3, 3′ diaminobenzidine tetrahydrochloride (DAB) is a carcinogen that should be used safely. It is one of the major sensitive substrates for HRP (horseradish peroxidase). DAB is brown or amber in color and becomes insoluble and precipitates over the antigen-antibody site due to the presence of hydrogen peroxide and peroxidase.
Application
- DAB Substrate is a color sustrate for peroxidases and is used in a variety of applications:Immunohistocytochemistry
- In situ hybridization
- Western blot
- Dot blot
DAB substrate has been used in immunofluorescence and western blot.
Packaging
1 set containing 2 components.
Preparation Note
Working solution:
Storage conditions (working solution): 2 to 8 °C
For best results, store the working solution at 2 to 8 °C when not using.
- Determine the volume of substrate required for the development of peroxidase.
- Remove the DAB/metal concentrate (10x) from -15 to -25 °C storage.
Note: Do not bring to 15 to 25 °C. - Dilute the DAB/metal concentrate (10x) with the peroxide buffer to a 1x working solution. For example, if 5 ml of substrate are required, dilute 500 μl of the DAB/metal concentrate (10x) with 4.5 ml of the Peroxide buffer.
- Mix well.
- Add the 1x working solution to the tissue or blot until desired substrate development.
Note: The 1x solution is stable over many hours.
For best results, store the working solution at 2 to 8 °C when not using.
Storage conditions (working solution): 2 to 8 °C
For best results, store the working solution at 2 to 8 °C when not using.
Other Notes
For life science research only. Not for use in diagnostic procedures.
Kit Components Only
Product No.
Description
- DAB/metal concentrate, 25 ml 10x concentrated
- Peroxide Buffer, 250 ml
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 - Aquatic Chronic 2 - Carc. 1A - Carc. 1B - Flam. Liq. 2 Oral - Repr. 1B - Skin Sens. 1 Inhalation - STOT RE 2 - STOT SE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The developmental origin of zygodactyl feet and its possible loss in the evolution of Passeriformes.
Botelho J F, et al.
Proc. Royal Soc. Lond. B., 281(1788), 20140765-20140765 (2014)
Adrenergic Receptor Protocols (2000)
The developmental origin of zygodactyl feet and its possible loss in the evolution of Passeriformes.
João Francisco Botelho et al.
Proceedings. Biological sciences, 281(1788), 20140765-20140765 (2014-06-27)
The zygodactyl orientation of toes (digits II and III pointing forwards, digits I and IV pointing backwards) evolved independently in different extant bird taxa. To understand the origin of this trait in modern birds, we investigated the development of the
Pathobiology of Human Disease: A Dynamic Encyclopedia of Disease Mechanisms (2014)
Genetic deletion of Wdr13 improves the metabolic phenotype of Lepr db/db mice by modulating AP1 and PPARγ target genes.
Singh V P, et al.
Diabetologia, 58(2), 384-392 (2015)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service