XX1614705
Millipore Ground Joint Flask for Vacuum Filtration
2L, Conical bottom
Synonym(s):
Filtration apparatus, Ground joint flask, Vacuum filtration flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104909
eCl@ss:
32031613
NACRES:
NB.24
Recommended Products
material
borosilicate glass
technique(s)
vacuum filtration: suitable
flask capacity
2 L
application(s)
filtration
shipped in
ambient
Related Categories
General description
Millipore Glass Vacuum Filtration Accessory Product
Ground Joint Flask, 2L, Conical bottom
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.
Ground Joint Flask, 2L, Conical bottom
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.
This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service