MXQTRT001
Milliflex® Quantum Membrane Transfer Tool
For use with Milliflex® Quantum system
Synonym(s):
Milliflex® Quantum Membrane Transfer Tool
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116129
eCl@ss:
32160606
NACRES:
NB.24
Recommended Products
material
black polyoxymethylene housing
silicone seal
stainless steel body (1.4462)
stainless steel lever(s) (1.4462)
Quality Level
manufacturer/tradename
Milliflex®
input
water
sample(s) (final product)
sample(s) (in process)
sample(s) (raw material)
sample(s) (environmental)
application(s)
pharmaceutical
shipped in
ambient
Related Categories
General description
Milliflex® Quantum membrane transfer tool is an additional accessory for Milliflex® Quantum Rapid Detection System. Milliflex® Quantum System is a rapid fluorescence-based technology for fast quantitative detection of contaminants over a broad range of filterable matrices. Therefore, the Milliflex® Quantum system is a fast and reliable alternative for the detection of microorganisms. The Milliflex®Quantum membrane transfer tool should be used in a microbiological safety cabinet or laminar flow hood. Place the Milliflex® Quantum membrane transfer tool onto the pump, lock it and perform easily the staining step of the Milliflex® Quantum workflow.
Application
Milliflex® Quantum membrane transfer tool is an additional accessory for the Milliflex® Quantum Rapid Detection System to ensure the optimal membrane fluorescent staining. It finds application in bioburden testing and water monitoring of beer, bottled water, cosmetics, and Pharmaceutical Products Analysis
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service