Skip to Content
Merck
All Photos(1)

Key Documents

MABC576

Sigma-Aldrich

Anti-HOIL-1 Antibody, clone 2E2

clone 2E2, from mouse

Synonym(s):

RanBP-type and C3HC4-type zinc finger-containing protein 1, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, HOIL-1, Hepatitis B virus X-associated protein 4, RING finger protein 54, Ubiquitin-conjugating enzyme 7-interacting protein 3, HO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RBCK1(10616)

General description

RanBP-type and C3HC4-type zinc finger-containing protein 1 (UniProt Q9BYM8; also known as HOIL-1, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, RBCC protein interacting with PKC1, RING finger protein 54, Ubiquitin-conjugating enzyme 7-interacting protein 3) is encoded by the RBCK1 gene (also known as C20orf18, HOIL1, PBMEI, RBCK2, RNF54, UBCE7IP3, XAP3, XAP4, ZRANB4) (Gene ID 10616) in human. The linear ubiquitin assembly complex (LUBAC) is comprised of two RING-IBR-RING (RBR)-containing E3 ligases, HOIL-1L and HOIP. LUBAC conjugates a head-to-tail-linked linear polyubiquitin chain onto lysine residues on substrate proteins. Ovarian tumor deubiquitinating enzyme (DUB) with linear linkage specificity (OTULIN/Gumby/FAM105B) and cylindromatosis (CYLD) are two DUBs that catalyze the cleavage linear polyubiquitin chains. OTULIN and CYLD interact with the N-terminal PNGase/UBA or UBX (PUB) domain of HOIP and negatively regulate LUBAC-mediated cellular processes. LUBAC is reported to suppress IFN-dependent antiviral activity by mediating TRIM25 degradation and competing against TRIM25 for RIG-I interaction.

Specificity

Clone 2E2 reacts with isoform 1 (HOIL-1L) and isoform 2, but not isoform 3.

Immunogen

Epitope: Near C-terminus
GST-tagged recombinant protein corresponding to human HOIL-1 near the C-terminus.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-HOIL-1 Antibody, clone 2E2 is validated for use in Western Blotting, Immunocytochemistry for the detection of HOIL-1 .
Western Blotting Analysis: A representative lot detected HOIL-1 in HeLa, SH-SY5Y, and U20S cells (Kirisako, T., et al (2006). EMBO J. 25(20):48877-4887).
Western Blotting Analysis: A representative lot detected HOIL-1 in mouse liver extracts (Tokunaga, F., et al (2009). Nat. Cell Biol. 11(2):123-132).
Western Blotting Analysis: A representative lot detected HOIL-1 in MEFs (Takiuchi, T., et al (2014). Genes Cells. 19(3):254-272).
Immunocytochemistry Analysis: A representative lot detected HOIL-1 in HeLa cells with or without Sendai virus (SeV) infection (Inn, K.S., et al. (2011). Mol Cell. 41(3):354-365).

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected HOIL-1 in 10 µg of THP-1 cell lysate.

Target description

~57 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirotsugu Oda et al.
Frontiers in immunology, 10, 479-479 (2019-04-03)
Background: HOIP is the catalytic subunit of the linear ubiquitination chain assembly complex (LUBAC) that is essential for NF-κB signaling and thus proper innate and adaptive immunity. To date only one patient with HOIP deficiency has been reported with clinical
Stephen C Walsh et al.
Cell host & microbe, 30(12), 1671-1684 (2022-09-10)
Chlamydia trachomatis is the leading cause of sexually transmitted bacterial infections and a major threat to women's reproductive health in particular. This obligate intracellular pathogen resides and replicates within a cellular compartment termed an inclusion, where it is sheltered by
Ying Jin et al.
Oncogenesis, 13(1), 6-6 (2024-01-26)
Hypoxia-inducible factor 1 (HIF1) is critically important for driving angiogenesis and tumorigenesis. Linear ubiquitin chain assembly complex (LUBAC), the only known ubiquitin ligase capable of catalyzing protein linear ubiquitination to date, is implicated in cell signaling and associated with cancers.
Dulcemaria Hernandez et al.
mBio, 13(5), e0188822-e0188822 (2022-09-27)
The obligate intracellular protozoan pathogen Toxoplasma gondii infects a wide range of vertebrate hosts and frequently causes zoonotic infections in humans. Whereas infected immunocompetent individuals typically remain asymptomatic, toxoplasmosis in immunocompromised individuals can manifest as a severe, potentially lethal disease
Nadine Weinelt et al.
Cell death & disease, 15(1), 77-77 (2024-01-21)
Plasma membrane accumulation of phosphorylated mixed lineage kinase domain-like (MLKL) is a hallmark of necroptosis, leading to membrane rupture and inflammatory cell death. Pro-death functions of MLKL are tightly controlled by several checkpoints, including phosphorylation. Endo- and exocytosis limit MLKL

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service