Skip to Content
Merck
All Photos(1)

Key Documents

MAB3804

Sigma-Aldrich

Anti-53BP1 Antibody, clone BP18

ascites fluid, clone BP18, Chemicon®

Synonym(s):

p53 Binding Protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

BP18, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TP53BP1(7158)

Specificity

53BP1

Immunogen

Recombinant human 53BP1 proteins.

Application

Anti-53BP1 Antibody, clone BP18 is a Mouse Monoclonal Antibody for detection of 53BP1 also known as p53 Binding Protein 1 & has been validated in IF & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western blot. Recognizes a protein of >250 kD. Suggested dilution buffer is TBST. Suggested blocking buffer is 5% non-fat dry milk.

Immunocytochemistry

Immunoprecipitation. Expected size in immunoblot is >250 kD. Suggested lysis buffer is NETN. Suggested capture agent is Protein A + rabbit anti-mouse linker antibody.

Optimal working dilutions must be determined by the end user.

Physical form

Ascites. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia E Rübe et al.
NPJ aging and mechanisms of disease, 7(1), 7-7 (2021-04-03)
Cellular senescence is an irreversible growth arrest that occurs as a result of damaging stimuli, including DNA damage and/or telomere shortening. Here, we investigate histone variant H2A.J as a new biomarker to detect senescent cells during human skin aging. Skin
Dejie Wang et al.
Science advances, 7(25) (2021-06-20)
53BP1 activates nonhomologous end joining (NHEJ) and inhibits homologous recombination (HR) repair of DNA double-strand breaks (DSBs). Dissociation of 53BP1 from DSBs and consequent activation of HR, a less error-prone pathway than NHEJ, helps maintain genome integrity during DNA replication;
Ishita Ghosh et al.
Nucleic acids research, 51(16), 8643-8662 (2023-07-13)
Environmental agents like ionizing radiation (IR) and chemotherapeutic drugs can cause severe damage to the DNA, often in the form of double-strand breaks (DSBs). Remaining unrepaired, DSBs can lead to chromosomal rearrangements, and cell death. One major error-free pathway to
Elisa Varela et al.
Nature communications, 7, 11739-11739 (2016-06-03)
Although telomere length is genetically determined, mouse embryonic stem (ES) cells with telomeres of twice the normal size have been generated. Here, we use such ES cells with 'hyper-long' telomeres, which also express green fluorescent protein (GFP), to generate chimaeric
Deeksha Munnur et al.
Cell reports, 26(8), 2028-2036 (2019-02-21)
Although poly-ADP-ribosylation (PARylation) of DNA repair factors had been well documented, its role in the repair of DNA double-strand breaks (DSBs) is poorly understood. NR4A nuclear orphan receptors were previously linked to DSB repair; however, their function in the process

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service