Skip to Content
Merck
All Photos(1)

Documents

IPFL20200

Millipore

Immobilon® -FL PVDF Membrane

10 sheets, 20cm x 20 cm, 0.45 µm pore size, transfer membrane with low background fluorescence

Synonym(s):

Western blotting membrane, blotting membrane, transfer membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
eCl@ss:
32031602
NACRES:
NB.22

product name

Immobilon®-FL PVDF Membrane, 10 sheets, 20 cm x 20 cm, 0.45 µm pore size, Hydrophobic PVDF Transfer Membrane with low background fluorescence for Western blotting. Compatible with visible and infrared fluorescent probes.

material

PVDF membrane
plain filter
white filter

Quality Level

feature

hydrophobic

manufacturer/tradename

Immobilon®

technique(s)

dot blot: suitable
western blot: suitable

filter L × W

20 cm × 20 cm

pore size

0.45 μm pore size

capacity

155 μg/cm2 adsorption capacity (insulin)
205 μg/cm2 adsorption capacity (BSA)
300 μg/cm2 adsorption capacity (goat IgG)

compatibility

for use with Amido black
for use with CPTS
for use with Coomassie brilliant blue
for use with Ponceau-S red

detection method

chemiluminescent
colorimetric
fluorometric

shipped in

ambient

General description

The Immobilon®-FL transfer membrane has a nominal pore size of 0.45 μm and is made up of polyvinylidene fluoride (PVDF) microporous membrane for binding proteins transferred from a variety of gel matrices. This membrane is hydrophobic and offers a uniformly controlled pore structure with a high binding capacity for biiomolecules. The membrane exhibits very low autofluorescence across a wide range of excitation/emission wavelengths. This property makes it ideal for blotting applications involving fluorescence-based immunodetection. In addition, the Immobilon®-FL membrane can be used for standard chemiluminescent or chromogenic detection. The Immobilon®-FL is recommended for optimal blotting of proteins with molecular weights greater than 20,000.

Application

Immobilon®-FL PVDF Membrane has been used in western blotting.

Features and Benefits

Immobilon®-FL Membrane:
  • Designed for optimal fluorescent immunodetection
  • Extremely low background improves the sensitivity of all fluorescence detection protocols
  • suitable for all commonly used fluorescent probes at all excitation and emission wavelengths
  • compatible with standard blocking agents and buffers
  • Ideal for multiplexing and chemifluorescence detection

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaoqiu Jiang et al.
Cell cycle (Georgetown, Tex.), 18(11), 1241-1253 (2019-05-14)
Researches have been focusing on the role of Slit2 in angiogenesis, specifically in cell migration and vessel permeability. Nevertheless, the role of Slit2-N, the bioactive fragment of Slit2, in the proliferation of vascular endothelia in choroidal neovascularization and some related
J L Portis et al.
Journal of virology, 83(12), 6079-6086 (2009-04-03)
A small group of ecotropic murine retroviruses cause a spongiform neurodegenerative disease manifested by tremor, paralysis, and wasting. The neurovirulence of these viruses has long been known to be determined by the sequence of the viral envelope protein, although the
Camilla Nehammer et al.
eLife, 8 (2019-12-05)
Appropriate regulation of autophagy is crucial for clearing toxic proteins from cells. Defective autophagy results in accumulation of toxic protein aggregates that detrimentally affect cellular function and organismal survival. Here, we report that the microRNA miR-1 regulates the autophagy pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service