HX0300
Hexane, mixture of isomers
≥98% (total hexane isomers + methylcyclopentane)
Synonym(s):
Hexanes
About This Item
Recommended Products
Product Name
Hexanes, 98%, Practical
vapor pressure
160 hPa ( 20 °C)
Quality Level
Assay
≥98% (total hexane isomers + methylcyclopentane)
form
liquid
color
APHA: ≤20
bp
69 °C/1013 hPa
mp
-94.3 °C
transition temp
flash point -22 °C
solubility
0.0095 g/L
density
0.66 g/cm3 at 20 °C
shipped in
ambient
storage temp.
room temp
SMILES string
C(CCC)CC
InChI
1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
InChI key
VLKZOEOYAKHREP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Packaging
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!
Analysis Note
Assay (total hexane isomers + methylcyclopentane): 98% min
Color (APHA): 20 max
Form: Clear liquid
Infrared Spectrum: Conforms
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system, Nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-7.6 °F - closed cup
Flash Point(C)
-22 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service