CDRM4HN05
Lynx® CDR
Lynx® CDR Male 1/2 Inch Hose Barb Reconnectable Connector - 5pack
Synonym(s):
Multiple aseptic connections, disconnections and reconnections (up to 6 times)
About This Item
Recommended Products
material
316 stainless steel spring(s) (electropolished, flow path)
polysulfone flow path (glass filled )
polysulfone valve body (glass filled)
silicone seal (flow path)
Quality Level
sterility
non-sterile
product line
Lynx®
parameter
0.7 bar max. pressure (10 psi) (connect/disconnect pressure)
2-40 °C operating temperature (36-104 °F)
4.1 bar max. pressure (60 psi) (operating pressure)
H
13.7 cm (5.4 in.)
L
25.7 cm (10.1 in.)
W
8.4 cm (3.3 in.)
fitting
1/2 in. inlet hose barb
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service