Skip to Content
Merck
All Photos(2)

Documents

ABT42

Sigma-Aldrich

Anti-Myosin-VI Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Myosin-VI, Unconventional myosin-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

canine, human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYO6(4646)

General description

Myosin-VI has been found in the membrane ruffles at the leading edge of the Golgi complex. Myosin-VI moves along the actin in the opposite way of other known myosins to date, toward the minus end. It can exist as a dimer and a monomer and is thought to function as a non-processive motor with a 18nm working stroke. Myosin-VI has also been found in the brain, specifically the synapses. Myosin-VI deficiency (Snell’s waltzer disease) in the hippocampus causes a smaller number of synapses, short dendritic spines and astrogliosis. Myosin-VI may also be important in clathrin-mediated endocytosis of acid-type glutamate receptors. Recent research has implicated myosin-VI in the enrichment of proteins on the surface of axons.

Immunogen

KLH-conjugated linear peptide corresponding to human Myosin-VI.

Application

Anti-Myosin-VI Antibody detects level of Myosin-VI & has been published & validated for use in WB & IC.
Immunocytochemistry analysis: A 1:500 dilution from a representative lot detected Myosin-VI in NIH/3T3 and A431 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in MDCK cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Myosin-VI in 10 µg of MDCK cell lysate.

Target description

~150 kDa observed.
An uncharacterized band appears at ~55 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MDCK cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service