Skip to Content
Merck
All Photos(2)

Key Documents

ABT121

Sigma-Aldrich

Anti-T-cadherin (CDH13) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cadherin-13, Heart cadherin, H-cadherin, P105, Truncated cadherin, T-cad, T-cadherin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
IHC
WB
species reactivity:
horse, human, rat
technique(s):
immunohistochemistry: suitable (paraffin)
western blot: suitable
citations:
8

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

horse, human, rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), equine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CDH13(1012)

General description

T-cadherin (CDH13), also known as Cadherin 13, is a member of the cadherin superfamily. This protein is unique from its other family members in that it lacks the cytoplasmic domains and is anchored to the cells membrane through the GPI anchor. T-cadherin is widely expressed in the cardiovascular system. Mutations in T-cadherin have been associated with many types of cancer.

Specificity

This antibody recognizes T-cadherin at the Cadherin 1 domain. This antibody detects the ~130 kDa pro-protein and the proteolytically cleaved ~105 kDa mature isoforms. (Denzel, M., et al. (2010). J Clin Invest.120(12):4342–4352.)

Immunogen

Epitope: Cadherin 1 domain
KLH-conjugated linear peptide corresponding to human T-cadherin at the Cadherin 1 domain.

Application

Anti-T-cadherin (CDH13) Antibody detects level of T-cadherin (CDH13) & has been published & validated for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected T-cadherin in human brain and human heart tissues.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Quality

Evaluated by Western Blot in human heart tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected T-cadherin on 10 µg of human heart tissue lysate.

Target description

~130 and ~105 kDa observed.
~130 kDa corresponds to the pro-protein and ~105 kDa corresponds to the proteolytically cleaved mature isoform. (Denzel, M., et al. (2010). J Clin Invest.120(12):4342–4352.)

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human heart tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhanya Dhanyalayam et al.
Frontiers in cardiovascular medicine, 9, 783974-783974 (2022-04-05)
Coronavirus disease-2019 (COVID-19) caused by Severe Acute Respiratory Syndrome Coronavirus 2 (SARS-CoV-2; CoV2) is a deadly contagious infectious disease. For those who survive COVID-19, post-COVID cardiac damage greatly increases the risk of cardiomyopathy and heart failure. Currently, the number of
Ken Okamura et al.
Pigment cell & melanoma research, 33(6), 826-833 (2020-06-20)
Racemic RS-4-(4-hydroxyphenyl)-2-butanol (rhododendrol; trade name: Rhododenol [RD]), which is used in topical skin-lightening cosmetics, was unexpectedly reported in Japan to induce leukoderma or vitiligo called RD-induced leukoderma (RIL) after repeated application. To our knowledge, no studies have investigated chemical-induced vitiligo
Weiwen Liu et al.
Journal of Cancer, 9(1), 117-128 (2018-01-02)
Dapivirine is one of reverse transcriptase inhibitors (RTIs). It is the prototype of diarylpyrimidines (DAPY), formerly known as TMC120 or DAPY R147681 (IUPAC name: 4- [[4-(2, 4, 6-trimethylphenyl) amino]-2-pyrimidinyl] amino]-benzonitrile; CAS no.244767-67-7). The purpose of this study is to investigate
Daniel J Tyrrell et al.
Arteriosclerosis, thrombosis, and vascular biology, 42(8), 1060-1076 (2022-05-06)
Aging enhances most chronic diseases but its impact on human aortic tissue in health and in thoracic aortic aneurysms (TAA) remains unclear. We employed a human aortic biorepository of healthy specimens (n=17) and those that underwent surgical repair for TAA
Julien Guerrero et al.
Stem cells translational medicine, 11(2), 213-229 (2022-03-09)
Cells of the stromal vascular fraction (SVF) of human adipose tissue have the capacity to generate osteogenic grafts with intrinsic vasculogenic properties. However, cultured adipose-derived stromal cells (ASCs), even after minimal monolayer expansion, lose osteogenic capacity in vivo. Communication between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service