Skip to Content
Merck
All Photos(1)

Key Documents

AB5320C3

Sigma-Aldrich

Anti-NG2 Chondroitin Sulfate Proteoglycan, Cy3 Conjugate Antibody

from rabbit, CY3 conjugate

Synonym(s):

Chondroitin sulfate proteoglycan 4, Chondroitin sulfate proteoglycan NG2, Melanoma chondroitin sulfate proteoglycan, Melanoma-associated chondroitin sulfate proteoglycan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

CY3 conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), monkey (based on 100% sequence homology), human (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CSPG4(1464)
mouse ... Cspg4(121021)
rat ... Cspg4(81651)
rhesus monkey ... Cspg4(713086)

General description

NG2 is a high molecular weight, integral membrane chondroitin sulfate proteoglycan. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells (i.e., O-2A progenitor cells). NG2 is also found on the surfaces of chondroblasts, proliferating capillary endothelial cells, some human melanoma cell lines, and on leukemic blasts in childhood acute lymphoblastic leukemia. The NG2 proteoglycan is likely to play a role in regulation of cell motility, axon outgrowth and the cellular responses to platelet-derived growth factor.

Specificity

This antibody identifies the intact proteoglycan and core protein by WB and ELISA. When oligodendrocyte precursor cells (i.e. O-2A progenitor cells) are stained alive, the stain appears as clusters on the cell surface. This antibody does not stain differentiated oligodendrocytes well.

Immunogen

Immunoaffinity purified NG2 Chondroitin Sulfate Proteoglycan from rat

Application

Anti-NG2 Chondroitin Sulfate Proteoglycan, Cy3 Conjugate Antibody is an antibody against NG2 Chondroitin Sulfate Proteoglycan for use in ICC & IHC.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Immunocytochemistry in rat oligodendrocyte precursor cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected NG2 in rat oligodendrocyte precursor cells. Performed without Triton.

Target description

The unconjugated parent antibody (Cat. No. AB5320) has an observed molecular weight at ~270-300 kDa

Physical form

Protein A purified
Purified rabbit polyclonal conjugated to Cy3 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Rat or Human oligodendrocyte precursor cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Sun et al.
Science translational medicine, 13(604) (2021-07-30)
The immature and dysfunctional vascular network within solid tumors poses a substantial obstacle to immunotherapy because it creates a hypoxic tumor microenvironment that actively limits immune cell infiltration. The molecular basis underpinning this vascular dysfunction is not fully understood. Using
Nadine Reichhart et al.
Diabetologia, 60(1), 202-211 (2016-10-06)
Although the renin-angiotensin system plays an important role in the progression of diabetic retinopathy, its influence therein has not been systematically evaluated. Here we test the suitability of a new translational model of diabetic retinopathy, the TetO rat, for addressing
Daniel Bormann et al.
Nature communications, 15(1), 6232-6232 (2024-07-24)
Neuroglia critically shape the brain´s response to ischemic stroke. However, their phenotypic heterogeneity impedes a holistic understanding of the cellular composition of the early ischemic lesion. Here we present a single cell resolution transcriptomics dataset of the brain´s acute response
Tamás Kovács-Öller et al.
International journal of molecular sciences, 21(7) (2020-04-09)
The nervous system demands an adequate oxygen and metabolite exchange, making pericytes (PCs), the only vasoactive cells on the capillaries, essential to neural function. Loss of PCs is a hallmark of multiple diseases, including diabetes, Alzheimer's, amyotrophic lateral sclerosis (ALS)
Elena Ivanova et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(32), 7580-7594 (2017-07-05)
Adequate blood flow is essential to brain function, and its disruption is an early indicator in diseases, such as stroke and diabetes. However, the mechanisms contributing to this impairment remain unclear. To address this gap, in the diabetic and nondiabetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service