Skip to Content
Merck
All Photos(1)

Key Documents

8.41034

Sigma-Aldrich

Aminoacetonitrile sulfate

for synthesis

Synonym(s):

Aminoacetonitrile sulfate, Glycinonitrile sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10N4O4S
CAS Number:
Molecular Weight:
210.21
MDL number:
UNSPSC Code:
12352107
EC Index Number:
226-772-0
NACRES:
NA.22
form:
powder
Assay:
≥96.0% (acidimetric)

Quality Level

Assay

≥96.0% (acidimetric)

form

powder

mp

118-122 °C

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(O)O.NCC#N.NCC#N

InChI

1S/2C2H4N2.H2O4S/c2*3-1-2-4;1-5(2,3)4/h2*1,3H2;(H2,1,2,3,4)

InChI key

NQHWLBZZBSYIPI-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 96.0 %
Identity (IR): passes test

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service