8.18707
Citric acid
anhydrous for synthesis
Synonym(s):
2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
About This Item
vapor pressure
<0.1 hPa ( 20 °C)
Quality Level
Assay
≥99% (alkalimetric)
form
solid
potency
3000 mg/kg LD50, oral (Rat)
expl. lim.
8 %, 65 °F
pH
1.7 (20 °C, 100 g/L in H2O)
pKa
(1) 3.13, (2) 4.76, (3) 6.4
bp
200 °C/1013 hPa (decomposition)
mp
153-159 °C (lit.)
solubility
1330 g/L
density
1.665 g/cm3 at 18 °C
bulk density
560 kg/m3
storage temp.
15-25°C
SMILES string
OC(=O)CC(O)(CC(O)=O)C(O)=O
InChI
1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)
InChI key
KRKNYBCHXYNGOX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Citric acid′s role in nanotechnology: Research on the use of citric acid as a precursor in the synthesis of carbon dots, highlighting its role in the creation of fluorescent nanoparticles for advanced material applications (J Schneider et al., 2017).
Analysis Note
Identity (IR-spectrum): passes test
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service