8.02809
2-Chloropropane
for synthesis
Synonym(s):
2-Chloropropane, Isopropyl chloride
About This Item
vapor pressure
592.1 hPa ( 20 °C)
Quality Level
form
liquid
autoignition temp.
590 °C
potency
>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
expl. lim.
2.8-10.7 % (v/v)
mp
-118 °C
transition temp
flash point -36 °C
density
0.86 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
ClC(C)C
InChI
1S/C3H7Cl/c1-3(2)4/h3H,1-2H3
InChI key
ULYZAYCEDJDHCC-UHFFFAOYSA-N
Application
- Development and validation of Genotoxic Impurities Chloromethane, Chloroethane and 2-Chloropropane, in Pregabalin drug substance using Head-Space: This study focuses on the detection and quantification of 2-Chloropropane as a genotoxic impurity in pharmaceutical products using head-space gas chromatography (P Bharath, PVS Gupta, SM Ali, D Ramachandran).
- Photocatalytic breakdown of 2-chloropropane: Research on the effectiveness of a photocatalytic reactor designed for the degradation of 2-Chloropropane in environmental applications (EC Ajiduku, E Kamba, R Odoh, 2023).
- Density and speed of sound of 2-chloropropane: An exploration into the physical properties of 2-Chloropropane under various temperatures and pressures, providing valuable data for industrial and laboratory applications (VV Melent′ev, EB Postnikov, 2019).
- Theoretic studies on the kinetics and mechanism of multi-channel gas-phase unimolecular reactions of 1-chloropropane and 2-chloropropane: Theoretical analysis providing insights into the thermal decomposition pathways of 2-Chloropropane, important for understanding its stability and reactivity (V Saheb, 2014).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-32.8 °F
Flash Point(C)
-36 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service