71281
ProteoJuice Protein Transfection Reagent
Efficient protein delivery into mammalian cells
Synonym(s):
Protein transfection reagent, Transfection reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106502
NACRES:
NA.54
Recommended Products
Quality Level
form
liquid
manufacturer/tradename
Novagen®
storage condition
OK to freeze
technique(s)
transfection: suitable
storage temp.
2-8°C
Related Categories
General description

<p>ProteoJuice<TMSYMBOL></TMSYMBOL> Protein Transfection Reagent is an effective reagent for the transfection of intact functional protein and peptides into mammalian cells with minimal toxicity and broad cell specificity. The chemistry of ProteoJuice differs substantially from protein delivery systems based on lipids or protein transduction domains. ProteoJuice forms a non-covalent complex with protein that protects against degradation by endosomal proteases and ensures delivery of intact protein inside the cell. Protein transfection with ProteoJuice is ideal for functional studies, delivering bioactive or inhibitory proteins, screening peptide libraries, studying subcellular location and turnover, and transient complementation of suppressed gene expression. Live cells can be examined less than two hours after delivery to determine the effects of the introduced material without the need for cell fixation.
<p>ProteoJuice Protein Transfection Reagent has been used successfully to deliver large proteins (such as antibodies), small proteins (such as histones), and peptides.


Efficient protein delivery into mammalian cells
Warning
Toxicity: Standard Handling (A)
Legal Information
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service