516485
Pepstatin A Methyl Ester
A cell-permeable methyl ester derivative of Pepstatin A that acts as a potent, non-competitive, transition-state analog inhibitor of γ-secretase.
Synonym(s):
Pepstatin A Methyl Ester, Isovaleryl-V V-Sta-A-Sta-OCH₃, PME
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C35H65N5O9
Molecular Weight:
699.92
UNSPSC Code:
12352108
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
lyophilized
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
solubility
DMSO: 5 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
A cell-permeable methyl ester derivative of Pepstatin A (Cat. No. 516481) that acts as a potent, non-competitive, transition-state analog inhibitor of γ-secretase (Kis = 150 nM, Kii = 320 nM in solubilized human γ-secretase at 20°C; Kis is the inhibition constant for inhibitor binding to the free enzyme and Kii is the inhibition constant for inhibitor binding to the Enzyme-Substrate complex). Sta = (3S,4S)-4-Amino-3-hydroxy-6-methyl-heptanoic acid.
A cell-permeable methyl ester derivative of Pepstatin A (Cat. No. 516482) that acts as a potent, non-competitive, transition-state analog inhibitor of γ-secretase (Kis = 15 nM and Kii = 32 nM in solubilized human γ-secretase; Kis is the inhibition constant for inhibitor binding to the free enzyme and Kii is the inhibition constant for inhibitor binding to the Enzyme-Substrate complex).
Biochem/physiol Actions
Cell permeable: yes
Primary Target
γ-secretase
γ-secretase
Product does not compete with ATP.
Reversible: no
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid freeze/thaw cycles of solutions.
Other Notes
Tian, G., et al. 2002. J. Biol. Chem.277, 31499.
Zhang, L., et al. 2001. Biochemistry40, 5049.
Murphy, M.P., et al. 1999. J. Biol. Chem.274, 11914.
Zhang, L., et al. 2001. Biochemistry40, 5049.
Murphy, M.P., et al. 1999. J. Biol. Chem.274, 11914.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service