Skip to Content
Merck
All Photos(1)

Key Documents

4410-OP

Millipore

Ethidium Bromide

OmniPur® Grade, Fluorescent stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105322
NACRES:
NA.28
Pricing and availability is not currently available.

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

dark red

foreign activity

DNAse, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

room temp

General description

Fluorescent stain for visualization of nucleic acids in gels. Facilitates purification of DNA when using a cesium chloride gradient. Molecular Biology grade.

Specifications
DNAse: none detected
RNAse: none detected
Protease: none detected

Warning

Toxicity: Toxic (F)

Physical form

Supplied as a 10 mg/ml solution in deionized water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Muta. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service