Skip to Content
Merck
All Photos(1)

Key Documents

420143

Sigma-Aldrich

JNK3 Inhibitor XII, SR-3576

The JNK3 Inhibitor XII, SR-3576, also referenced under CAS 1164153-22-3, controls the biological activity of JNK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK3 Inhibitor XII, SR-3576, 3-(4-(3-(3-Methylphenylureido))-1H-pyrazol-1-yl)-N-(3,4,5-trimethoxyphenyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27N5O5
CAS Number:
Molecular Weight:
501.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[n]3(ncc(c3)NC(=O)Nc4cc(ccc4)C)c1cc(ccc1)C(=O)Nc2cc(c(c(c2)OC)OC)OC

InChI

1S/C27H27N5O5/c1-17-7-5-9-19(11-17)30-27(34)31-21-15-28-32(16-21)22-10-6-8-18(12-22)26(33)29-20-13-23(35-2)25(37-4)24(14-20)36-3/h5-16H,1-4H3,(H,29,33)(H2,30,31,34)

InChI key

MTFAYLZZDJGFGV-UHFFFAOYSA-N

General description

A cell-permeable pyrazolourea compound that acts as a potent, ATP binding pocket-targeting, and JNK3-selective inhibitor (IC50 = 7 nM), inhibiting JNK1 only at much higher concentrations (IC50 = 170 nM) and exhibiting little activity against p38 even at concentrations as high as 20 µM. Shown to inhibit streptozotocin- (Cat. No. 572201) induced c-jun phosphorylation in INS-1 rat β-pancreatic cells (IC50 = 1.3 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kamenecka, T., et al. 2009. J. Biol. Chem.284, 12853.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service