Skip to Content
Merck
All Photos(1)

Documents

17-10393

Sigma-Aldrich

Rac1 Activation Magnetic Beads Pulldown Assay

Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Rac1-GTP from cell lysates, & detection by a Rac1 specific monoclonal antibody.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (G-protein activation)
affinity binding assay: suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... RAC1(5879)

Specificity

Predicted to cross-react with all mammalian species
Anti-Rac1, clone 23A8 (mouse monoclonal IgG2b): Human, mouse and rat, other species cross-reactivity unknown. Note: This antibody may be reused 3-5 times for immunoblot analysis. Add 0.05% sodium azide to the blocking buffer containing the diluted antibody. Do not use sodium azides with HRP

Application

Non-radioactive Rac1 Activation Assay Kit can be used to precipitate Rac1-GTP from cell lysates, & detection by a Rac1 specific monoclonal antibody.
Research Category
Signaling

Packaging

Kit capacity: 30 assays

Components

Rac1/Cdc42 Assay Reagent (Pak-1 PBD, Magnetic beads): 300 ug of Pak-1 PBD in 150 uL of glutathione magnetic beads, provided as a 50% beads slurry in PBS containing 50% glycerol for a final volume of 300 uL.

Anti-Rac1, clone 23A8: (Part No. 05-389) One vial containing 250 ug of purified IgG2akappa in 250 uL of storage buffer (0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, containing 0.05% sodium azide).

Mg2+ Lysis/Wash Buffer, 5X, (Part No. 20-168).Two vials, each vial contains 18 mL of 125 mM, HEPES, pH 7.5, 750 mM NaCl, 5% Igepal CA-630, 50mM MgCl2, 5 mM EDTA and 10% glycerol

100X GTPγS, 10mM (Cat.# 20-176) One vial containing 50 uL of 10 mM GTPγS, 100X stock, in sterile water. Non-hydrolyzable analog of GTP. Sufficient to label 5 mL of cell lysates

100X GDP, 100 mM, (Part No. 20-177). One vial containing 50 uL of 100 mM GDP, 100X stock, in sterile water. GDP, Guanosine 5’-Diphosphate, type I, for in vitro labeling of G-proteins in the inactive form. Sufficient to label 5ml of cell lysates.

Quality

Routinely evaluated by precipitating Rac-GTP from 3T3 cell lysates that had been loaded with GTPγS. The precipitated Rac-GTP was detected by immunoblot analysis using anti-Rac.

Linkage

Replaces: SGT425

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Yi Yang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(25), 4959-4975 (2019-04-18)
Coiled-coil and C2 domain containing 1A (CC2D1A) is an evolutionarily conserved protein, originally identified as a nuclear factor-κB activator through a large-scale screen of human genes. Mutations in the human Cc2d1a gene result in autosomal recessive nonsyndromic intellectual disability. It
Samuel J Atkinson et al.
EMBO reports, 19(7) (2018-05-26)
Integrin β3 is seen as a key anti-angiogenic target for cancer treatment due to its expression on neovasculature, but the role it plays in the process is complex; whether it is pro- or anti-angiogenic depends on the context in which
Kitti Szabo et al.
Cellular and molecular life sciences : CMLS, 79(2), 122-122 (2022-02-08)
Skeletal muscle demonstrates a high degree of regenerative capacity repeating the embryonic myogenic program under strict control. Rhabdomyosarcoma is the most common sarcoma in childhood and is characterized by impaired muscle differentiation. In this study, we observed that silencing the expression
Wensheng Liu et al.
Oncotarget, 6(11), 8851-8874 (2015-04-11)
N-myc downstream regulated gene-1 (NDRG1) is a potent metastasis suppressor that plays a key role in regulating signaling pathways involved in mediating cancer cell invasion and migration, including those derived from prostate, colon, etc. However, the mechanisms and molecular targets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service