1.19208
Silver oxide 99+
for analysis 99+ EMSURE®
Synonym(s):
Silver oxide 99+
About This Item
Recommended Products
Quality Level
product line
EMSURE®
Assay
99.0-101.0% Ag basis
form
powder
potency
2820 mg/kg LD50, oral (Rat)
mp
>200 °C (decomposes)
density
7.2 g/cm3 at 20 °C
bulk density
950 kg/m3
cation traces
Ag: 92.2-94.0%
Al: ≤10 ppm
Ca: ≤50 ppm
Cd: ≤10 ppm
Co: ≤10 ppm
Cr: ≤10 ppm
Cu: ≤50 ppm
Fe: ≤50 ppm
Hg: ≤100 ppm
Ir: ≤100 ppm
K: ≤50 ppm
Mg: ≤50 ppm
Mn: ≤10 ppm
Na: ≤100 ppm
Ni: ≤10 ppm
Os: ≤100 ppm
Pb: ≤50 ppm
Pd: ≤100 ppm
Pt: ≤100 ppm
Rh: ≤100 ppm
Ru: ≤100 ppm
Zn: ≤10 ppm
storage temp.
2-8°C
InChI
1S/2Ag.H2O/h;;1H2
InChI key
VFWRGKJLLYDFBY-UHFFFAOYSA-N
Related Categories
Analysis Note
Assay (calculated of Ag): 99.0 - 101.0 %
Ag (Silver): 92.2 - 94.0 %
Al (Aluminium): ≤ 10 ppm
Ca (Calcium): ≤ 50 ppm
Cd (Cadmium): ≤ 10 ppm
Co (Cobalt): ≤ 10 ppm
Cr (Chromium): ≤ 10 ppm
Cu (Copper): ≤ 50 ppm
Fe (Iron): ≤ 50 ppm
Hg (Mercury): ≤ 100 ppm
Ir (Iridium): ≤ 100 ppm
K (Potassium): ≤ 50 ppm
Mg (Magnesium): ≤ 50 ppm
Mn (Manganese): ≤ 10 ppm
Na (Sodium): ≤ 100 ppm
Ni (Nickel): ≤ 10 ppm
Os (Osmium): ≤ 100 ppm
Pb (Lead): ≤ 50 ppm
Pd (Palladium): ≤ 100 ppm
Pt (Platinum): ≤ 100 ppm
Rh (Rhodium): ≤ 100 ppm
Ru (Ruthenium): ≤ 100 ppm
Zn (Zinc): ≤ 10 ppm
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service