Skip to Content
Merck
All Photos(5)

Key Documents

1.08293

Supelco

Cyclopentyl methyl ether

greener alternative

EMPLURA®

Synonym(s):

CPME

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
MDL number:
UNSPSC Code:
12352112
NACRES:
NA.05
bp:
106 °C/760 mmHg
vapor pressure:
59.9 hPa ( 25 °C)
Pricing and availability is not currently available.

vapor pressure

59.9 hPa ( 25 °C)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

potency

200-2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

dilution

(low-cost alternative, for basic applications)

bp

106 °C/760 mmHg

mp

-140 °C

transition temp

flash point -1 °C

density

0.86 g/mL at 25 °C

greener alternative category

storage temp.

2-25°C

SMILES string

COC1CCCC1

InChI

1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3

InChI key

SKTCDJAMAYNROS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Cyclopentyl methyl ether (CPME) is a greener alternative to other ethereal solvents such as tetrahydrofuran (THF), 2-methyl tetrahydrofuran (2-MeTHF), dioxane (carcinogenic), tert-Butyl methyl ether (MTBE), and 1,2-dimethoxyethane (DME) and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information.

Application


  • Investigating the efficacy of green solvents and solvent-free conditions in hydrogen-bonding mediated organocatalyzed model reactions.: This study explores alternative green solvents, including Cyclopentyl methyl ether, assessing their effectiveness in catalyzing organic reactions under environmentally friendly conditions (Govender et al., 2024).

  • A systematic investigation of sample diluents in modern supercritical fluid chromatography.: Research focused on the impact of different solvents, such as Cyclopentyl methyl ether, on the performance of supercritical fluid chromatography, highlighting improvements in analytical accuracy and solvent efficiency (Guillarme et al., 2017).

  • Evaluation of non-conventional polar modifiers on immobilized chiral stationary phases for improved resolution of enantiomers by supercritical fluid chromatography.: This publication discusses the role of Cyclopentyl methyl ether as a polar modifier in chromatographic methods, enhancing the chiral separation capabilities in pharmaceutical analysis (Welch et al., 2014).


Features and Benefits

Greener alternative for THF, diethyl ether, and MTBE

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

< 3 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service