Skip to Content
Merck
All Photos(1)

Key Documents

1.07961

Sigma-Aldrich

Entellan new

rapid mounting medium for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401
NACRES:
NA.21
Pricing and availability is not currently available.

vapor pressure

<8 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

>250 °C

expl. lim.

1.1-8.0 % (v/v) xylene()

IVD

for in vitro diagnostic use

color

colorless

refractive index

n20/D 1.490-1.500

bp

137-143 °C/1013 hPa

transition temp

flash point 23 °C

density

0.95 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Entellan new - rapid mounting medium for microscopy, is a ready-to-use, water-free mounting medium for microscopy that consists of a polymer of mixed acrylates which are solubilized in xylene. Therefore, it should be used with specimens that have been cleared with xylene previous mounting. Its narrow viscosity range improves and accelerates the application, as neither the dropping rate nor the volume need to be reset after the first run with a new bottle of Entellan new. Entellan new is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Refractive index (20°C) 1.490 - 1.500
Density (20°C / 4°C) 0.94 - 0.96 g/cm3
Viscosity (20°C) 250 - 600 mPa*s

Analysis Note

Refractive index (n 20/D): 1.490 - 1.500
Density (d 20 °C/ 4 °C): 0.94 - 0.96
Fluorescence (as quinine at 365 nm): ≤ 250 ppb
Viscosity (20 °C): 250 - 600 mPa∙s
Suitability for microscopy: passes test

Legal Information

ENTELLAN is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samara Damasceno et al.
Frontiers in neurology, 11, 33-33 (2020-03-03)
The Wistar Audiogenic Rat (WAR) and the Genetic Audiogenic Seizure Hamster from Salamanca (GASH/Sal) strains are audiogenic epilepsy models, in which seizures are triggered by acoustic stimulation. These strains were developed by selective reproduction and have a genetic background with
Sandra Barata-Antunes et al.
International journal of molecular sciences, 21(10) (2020-05-20)
Parkinson's disease (PD) is the second most common age-related neurodegenerative disorder. The neurodegeneration leading to incapacitating motor abnormalities mainly occurs in the nigrostriatal pathway due to the loss of dopaminergic neurons in the substantia nigra pars compacta (SNpc). Several animal
Thomas Bise et al.
Scientific reports, 10(1), 11551-11551 (2020-07-16)
Zebrafish can regenerate their damaged hearts throughout their lifespan. It is, however, unknown, whether regeneration remains effective when challenged with successive cycles of cardiac damage in the same animals. Here, we assessed ventricular restoration after two, three and six cryoinjuries
Matthias Schnetz et al.
Cancers, 12(3) (2020-02-29)
Accumulating evidence suggests that iron homeostasis is disturbed in tumors. We aimed at clarifying the distribution of iron in renal cell carcinoma (RCC). Considering the pivotal role of macrophages for iron homeostasis and their association with poor clinical outcome, we

Questions

  1. Which other clearing agents is Entellan New compatible with apart from Xylene?

    1 answer
    1. It is recommended to use a mounting media like NeoClear, which is formulated with a xylene substitute. Toluene or Dioxane could be used for the same purposes

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service