Skip to Content
Merck
All Photos(1)

Documents

1.05653

Supelco

Lithium nitrate

99.995 Suprapur®

Synonym(s):

Lithium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiNO3
CAS Number:
Molecular Weight:
68.95
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-218-9
NACRES:
NA.21

grade

for inorganic trace analysis

Quality Level

Assay

99.995%

form

solid

pH

7-9 (20 °C, 50 g/L in H2O)

mp

255 °C

solubility

522 g/L

density

2.36 g/cm3 at 20 °C

bulk density

910 kg/m3

storage temp.

2-30°C

InChI

1S/Li.NO3/c;2-1(3)4/q+1;-1

InChI key

IIPYXGDZVMZOAP-UHFFFAOYSA-N

Application

  • Catalytic Behavior of Lithium Nitrate in Li-O2 Cells.: This study explores the catalytic role of lithium nitrate in lithium-oxygen batteries, demonstrating its effectiveness in enhancing the performance and stability of Li-O2 cells. The research highlights lithium nitrate′s potential to improve battery efficiency by facilitating the oxygen reduction and evolution reactions, making it a valuable component in advanced battery technologies (Sharon et al., 2015).

Analysis Note

Purity (metallic): ≥ 99.995 %
Assay (perchloric acid titration): ≥ 99.0 %
Chloride (Cl): ≤ 10 ppm
Phosphate (PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 10 ppm
Al (Aluminium): ≤ 0.02 ppm
Ba (Barium): ≤ 1.0 ppm
Ca (Calcium): ≤ 1.0 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.005 ppm
Cs (Cesium): ≤ 5 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.005 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 5 ppm
La (Lanthanum): ≤ 0.005 ppm
Mg (Magnesium): ≤ 0.5 ppm
Mn (Manganese): ≤ 0.005 ppm
Na (Sodium): ≤ 5 ppm
Ni (Nickel): ≤ 0.005 ppm
Pb (Lead): ≤ 0.005 ppm
Rb (Rubidium): ≤ 0.5 ppm
Sc (Scandium): ≤ 0.005 ppm
Sm (Samarium): ≤ 0.005 ppm
Sr (Strontium): ≤ 1.0 ppm
Tl (Thallium): ≤ 0.010 ppm
Y (Yttrium): ≤ 0.005 ppm
Yb (Ytterbium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.020 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service