Skip to Content
Merck
All Photos(1)

Key Documents

1.04904

Supelco

Potassium bromide

99.999 Suprapur®

Synonym(s):

Potassium bromide, Hydrobromic Acid Potassium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BrK
CAS Number:
Molecular Weight:
119.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-830-3
NACRES:
NB.21
Assay:
99.999%
grade:
for inorganic trace analysis
form:
solid
solubility:
650 g/L

grade

for inorganic trace analysis

Quality Level

vapor pressure

1.3 hPa ( 795 °C)

Assay

99.999%

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

5.5-8.5 (20 °C, 50 g/L in H2O)

bp

1435 °C/1013 hPa

mp

730 °C

solubility

650 g/L

density

2.75 g/cm3 at 20 °C

bulk density

900‑1000 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Application


  • Development of a new continuous homogeneous liquid phase microextraction procedure based on in-situ preparation of deep eutectic solvent: This research introduces a novel method for the analysis of aliphatic amines in urine samples, which may include potassium bromide as a part of the matrix or reagents used in the GC-MS process, demonstrating its versatility in biochemical sampling and testing (Shagaghipour S et al., 2024).

  • Classification of asbestos and their nonasbestiform analogues using FTIR and multivariate data analysis: This study utilizes FTIR spectroscopy, potentially involving potassium bromide in IR spectroscopic applications, to classify hazardous materials, illustrating the compound′s critical role in environmental safety and industrial materials analysis (Lee T et al., 2024).

  • A toolbox for easy entry low wavenumber in situ atomic layer deposition transmission FTIR spectroscopy studies: Demonstrates the use of potassium bromide in the enhancement of FTIR spectroscopy, particularly beneficial for the material science field, providing a critical tool for in-depth material analysis (Bin Afif A et al., 2022).

  • Halogen ion-modified silver nanoparticles for ultrasensitive surface-enhanced Raman spectroscopy detection of polycyclic aromatic hydrocarbons: Explores the use of potassium bromide in modifying nanoparticles for enhanced Raman spectroscopy, underlining its significant contributions to chemical sensing and environmental monitoring (Wang D et al., 2022).


Analysis Note

Purity (metallic): ≥ 99.999 %
Assay (argentometric): ≥ 99.5 %
Bromate (BrO₃): ≤ 10 ppm
Chloride (Cl): ≤ 500 ppm
Iodide (I): ≤ 200 ppm
Sulfate (SO₄): ≤ 10 ppm
Total nitrogen (N): ≤ 2 ppm
Al (Aluminium): ≤ 0.02 ppm
Ba (Barium): ≤ 0.5 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.005 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.005 ppm
Cu (Copper): ≤ 0.005 ppm
Fe (Iron): ≤ 0.010 ppm
La (Lanthanum): ≤ 0.005 ppm
Li (Lithium): ≤ 0.5 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.005 ppm
Na (Sodium): ≤ 5.0 ppm
Ni (Nickel): ≤ 0.005 ppm
Pb (Lead): ≤ 0.005 ppm
Sc (Scandium): ≤ 0.005 ppm
Sm (Samarium): ≤ 0.005 ppm
Sr (Strontium): ≤ 0.05 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.005 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service