A 0.04% indicator solution of bromothymol blue sodium salt can be prepared by dissolving 0.2 grams of Product No. 1.01895 in 500 mL pure water.
1.01895
Bromothymol blue sodium salt
pH indicator, water-soluble, ACS
Synonym(s):
Bromothymol blue sodium salt
About This Item
Recommended Products
Product Name
Bromothymol blue sodium salt, indicator water-soluble ACS
grade
ACS reagent
Quality Level
loss
≤5.0% loss on drying, 110°C
visual transition interval
5.8-7.6, yellow to blue
storage temp.
2-30°C
SMILES string
[Na+].[S](=O)(=O)(O)c1c(cccc1)\C(=C3/C(=C(C(=O)C(=C/3)C(C)C)Br)C)\c2c(c(c(c(c2)C(C)C)[O-])Br)C
InChI
1S/C27H28Br2O5S.Na/c1-13(2)18-11-20(15(5)24(28)26(18)30)23(17-9-7-8-10-22(17)35(32,33)34)21-12-19(14(3)4)27(31)25(29)16(21)6;/h7-14,30H,1-6H3,(H,32,33,34);/q;+1/p-1/b23-21+;
InChI key
NMKFVGALBGZKGW-FKWCIMQXSA-M
Related Categories
Application
- Effect of small molecules on the phase behavior and coacervation of aqueous solutions of poly(diallyldimethylammonium chloride) and poly(sodium 4-styrene sulfonate).: This study explores how small molecules like Bromothymol blue sodium salt influence the phase behavior and coacervation in polyelectrolyte solutions. This insight is crucial for understanding complex coacervate systems used in various applications such as drug delivery and water treatment (Huang et al., 2018).
- A wearable fingernail chemical sensing platform: pH sensing at your fingertips.: The research introduces an innovative wearable sensor using Bromothymol blue sodium salt for pH detection. This platform is significant for real-time monitoring of chemical exposures, particularly useful in clinical and environmental settings (Kim et al., 2016).
- Use of urease-bromothymol blue-agar method for large-scale testing of urine on grain and seeds.: This paper discusses a method involving Bromothymol blue sodium salt for detecting urea in grain and seeds, which is vital for quality control in agriculture and food safety (Valdes et al., 1996).
- The mechanism of liposomal damage by taurocholate.: This study examines the damaging effects of taurocholate on liposomes using Bromothymol blue sodium salt as a pH indicator, contributing to the understanding of bile salt interactions with lipid membranes (Walde et al., 1987).
Analysis Note
Appearance of solution: conforms
Loss on drying (110 °C): ≤ 5.0 %
Transition range (according to ACS): conforms
ACS conforms
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How do I make a BTB solution from Bromothymol Blue Sodium salt
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service