Skip to Content
Merck
All Photos(1)

Key Documents

1.00608

Supelco

Chlorine Dioxide Test, photometric

photometric, 0.020-10.00 mg/L (ClO2), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Chlorine Dioxide Test, photometric, 0.020-10.00 mg/L (ClO2), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 200 tests

specific analyte(s)

chlorine

measuring range

0.020-10.00 mg/L (ClO2)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Move DC
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (DPD)

storage temp.

15-25°C

General description

Method: photometric, DPD 0.020 - 10.00 mg/l ClO₂ Spectroquant®
This Spectroquant® Chlorine dioxide test allows the accurate quantification in various waters. As one of several oxides of chlorine, it is a potent and useful oxidizing agent used in water treatment and in bleaching. In weakly acidic solution chlorine dioxide reacts with dipropyl-p-phenylenediamine (DPD) to form a red-violet dye that is determined photometrically. Chlorine is masked prior to the color reaction.
The method is analogous to APHA 4500-ClO2 D and DIN 38408-5.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Chlorine dioxide acc. to DIN EN ISO 7393

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service