Skip to Content
Merck
All Photos(3)

Documents

07-1436

Sigma-Aldrich

Anti-PDGF-AA Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Platelet-derived growth factor alpha, Platelet-derived growth factor alpha chain, Platelet-derived growth factor alpha polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
neutralization: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PDGFA(5154)

Related Categories

General description

Platelet-Derived Growth Factor (PDGF) is a dimer of A (17 kDa) and B (14 kDa) chains (AA, AB, or BB). Its receptors are transmembrane receptor tyrosine kinases, PDGF Receptors type A and B. The type A PDGF receptor binds all three PDGFs with high affinity, whereas the type B PDGF receptor is selective for PDGF-BB. PDGF signals as a growth and survival factor, as it is a potent activator of the PI3 Kinase pathway. PDGF signaling appears to play an essential role in the development of tumors of glial origin.

Specificity

Reacts with PDGF-AA

Immunogen

Highly pure (>98%) recombinant human PDGF-AA.

Application

Detect PDGF-AA using this Anti-PDGF-AA Antibody validated for use in IH(P), ELISA, WB, NEUT.
Immunohistochemisty (paraffin): PDGF representative staining pattern/morphology in prostate cancer. Tissue was pretreated with citrate pH 6, antigen retrieval. A previous lot of this antibody was diluted to 1:100, using IHC-Select Detection with HRP-DAB.

ELISA: 0.5 μg/mL dilution of a previous lot allowed detection (in direct ELISA) of 0.2-0.4 ng/well when human PDGF-AA is used to coat the plate.

Neutralization: To yield ½ maiximal inhibition (ND50) of the biological activity of hPDGF-AA (5.0 ng/mL), a concentration of 0.05-0.07 μg/mL of a previous lot of this antibody was required.

Optimal working dilutions must be determined by the end user.
Research Category
Signaling
Research Sub Category
Growth Factors & Receptors

Quality

Routinely evaluated by Western Blot on C2C12 lysates.

Western Blot Analysis:
1:500 dilution of this lot detected PDGF-AA on 10 μg of C2C12 lysates.

Target description

24 kDa detected in Western Blot.

Linkage

Replaces: AB1486P

Physical form

Affinity purified
Purified rabbit polyclonal IgG lyophilized from PBS, pH 7.2.

Storage and Stability

The lyophilized antibody is stable at room temperature for at least one month and for up to one year when kept at -20ºC. When reconstituted in sterile water to a concentration of 0.1-1.0 mg/mL, the antibody is stable for at least six weeks at 2-8ºC.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
C2C12 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka P Looney et al.
Bio-protocol, 9(16) (2019-09-19)
Pulmonary fibrosis is characterized by pathological scaring of the lung. Similar to other fibrotic diseases, scar formation is driven by excessive extracellular matrix deposition by activated, proliferative, and migratory fibroblasts. Currently, the two most widely used chemotaxis and cell migration
Activation of unfolded protein response in transgenic mouse lenses.
Reneker, LW; Chen, H; Overbeek, PA
Investigative Ophthalmology & Visual Science null
Neural grafts containing exosomes derived from Schwann cell-like cells promote peripheral nerve regeneration in rats.
Hu, et al.
Burns & trauma, 11, tkad013-tkad013 (2023)
Tianrun Liu et al.
Nature communications, 9(1), 3439-3439 (2018-08-29)
Angiogenesis is a hallmark of cancer. However, most malignant solid tumors exhibit robust resistance to current anti-angiogenic therapies that primarily target VEGF pathways. Here we report that endothelial-mesenchymal transformation induces glioblastoma (GBM) resistance to anti-angiogenic therapy by downregulating VEGFR-2 expression
Tamihiro Kamata et al.
Cell reports, 31(12), 107802-107802 (2020-06-25)
The tumor microenvironment (TME) consists of different cell types, including tumor-associated macrophages (TAMs) and tumor-associated fibroblasts (TAFs). How these cells interact and contribute to lung carcinogenesis remains elusive. Using G12DKRAS- and V600EBRAF-driven mouse lung models, we identify the pleiotropic glycoprotein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service