Skip to Content
Merck
All Photos(1)

Key Documents

P-004

Supelco

Phenobarbital-D5 (side chain) solution

(deuterium label on side chain), 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D5H7N2O3
CAS Number:
Molecular Weight:
237.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C1(C(=O)NC(=O)NC1=O)c2ccccc2

InChI

1S/C12H12N2O3/c1-2-12(8-6-4-3-5-7-8)9(15)13-11(17)14-10(12)16/h3-7H,2H2,1H3,(H2,13,14,15,16,17)/i1D3,2D2

InChI key

DDBREPKUVSBGFI-ZBJDZAJPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

An internal standard suitable for quantitation of phenobarbital levels in GC/MS or LC/MS applications for clinical toxicology, urine drug testing, or forensic analysis. Phenobarbital is a barbiturate used to treat all epileptic seizure types except for absence seizures. Phenobarbital is the oldest and most widely used anticonvulsant.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georg Petkau et al.
Nature communications, 13(1), 2274-2274 (2022-04-29)
CD8+ T cell differentiation into effector cells is initiated early after antigen encounter by signals from the T cell antigen receptor and costimulatory molecules. The molecular mechanisms that establish the timing and rate of differentiation however are not defined. Here
Eugenie Werbenko et al.
Scientific reports, 14(1), 8109-8109 (2024-04-07)
Bone resorption is highly dependent on the dynamic rearrangement of the osteoclast actin cytoskeleton to allow formation of actin rings and a functional ruffled border. Hem1 is a hematopoietic-specific subunit of the WAVE-complex which regulates actin polymerization and is crucial
Álvaro Santana-Garrido et al.
Molecular nutrition & food research, 68(3), e2200623-e2200623 (2023-12-04)
Our laboratory has previously described the antioxidant and anti-inflammatory potential of a wild olive (acebuche, ACE) oil against hypertension-associated vascular retinopathies. The current study aims to analyze the antifibrotic effect of ACE oil on the retina of hypertensive mice. Mice are
Takuya Ikeda et al.
Cell death discovery, 9(1), 454-454 (2023-12-15)
Nogo-Nogo receptor 1 (NgR1) signaling is significantly implicated in neurodegeneration in amyotrophic lateral sclerosis (ALS). We previously showed that lateral olfactory tract usher substance (LOTUS) is an endogenous antagonist of NgR1 that prevents all myelin-associated inhibitors (MAIs), including Nogo, from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service