Skip to Content
Merck
All Photos(1)

Key Documents

890700P

Avanti

12:0 EPC (Cl Salt)

Avanti Research - A Croda Brand 890700P, powder

Synonym(s):

1,2-dilauroyl-sn-glycero-3-ethylphosphocholine (chloride salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H69NO8PCl
CAS Number:
Molecular Weight:
686.34
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (890700P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890700P

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O)OCC.[Cl-]

InChI

1S/C34H69NO8P.ClH/c1-7-10-12-14-16-18-20-22-24-26-33(36)39-30-32(31-42-44(38,40-9-3)41-29-28-35(4,5)6)43-34(37)27-25-23-21-19-17-15-13-11-8-2;/h32H,7-31H2,1-6H3;1H/q+1;/p-1/t32-,44?;/m1./s1

InChI key

QWVQEMZYQQUQCH-XFSYIJCRSA-M

General description

1,2-dilauroyl-sn-glycero-3-ethylphosphocholine (12:0 EPC) is a cationic phospholipid. 12:0 EPC is a short-chain analog of dioleoyl ethylphosphatidylcholine. O-alkyl phosphatidylcholines constitute the first chemically stable triesters of biological lipid structures and the first cationic derivatives of phospholipids consisting entirely of biological metabolites linked with ester bonds. The lipid has low toxicity and is biodegradable.

Application

1,2-dilauroyl-sn-glycero-3-ethylphosphocholine (12:0 EPC (Cl Salt)) is suitable for use in lipoplex preparation and to study the lipoplex transfection activity. It might also be used in lipid mixtures to study its cubic phases of formation, stability and phase transitions.

Biochem/physiol Actions

1,2-dilauroyl-sn-glycero-3-ethylphosphocholine (12:0 EPC) is preferred for transfection of cultured primary endothelial cells. 12:0 EPC forms lamellar lipoplexes possessing high to superior transfection activity. Chemically synthesized cationic lipids are applicable as non-viral gene carriers. Variation in the hydrocarbon chain of phosphatidylcholine (PC) derivatives affects transfection efficiency.

Packaging

5 mL Clear Glass Sealed Ampule (890700P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Wang et al.
Biophysical journal, 91(10), 3692-3706 (2006-08-29)
A combination of two cationic lipid derivatives having the same headgroup but tails of different chain lengths has been shown to have considerably different transfection activity than do the separate molecules. Such findings point to the importance of investigating the
Analysis of lipoplex structure and lipid phase changes
Liposomes, 399-423 (2010)
Boris Tenchov et al.
Chemistry and physics of lipids, 208, 65-74 (2017-10-07)
The non-lamellar phases formed by membrane lipids in diluted aqueous dispersions are mainly represented by the inverted hexagonal phase, HII, and phases of cubic symmetry, among them the bicontinuous cubic phases Pn3m (Q224), Im3m (Q229) and Ia3d (Q230). Here we
Rumiana Koynova et al.
The journal of physical chemistry. B, 111(27), 7786-7795 (2007-06-19)
Some mixtures of two cationic lipids including phospholipid compounds (O-ethylphosphatidylcholines) as well as common, commercially available cationic lipids, such as dimethylammonium bromides and trimethylammonium propanes, deliver therapeutic DNA considerably more efficiently than do the separate molecules. In an effort to
Li Wang et al.
Molecular pharmaceutics, 4(4), 615-623 (2007-04-06)
To date, the primary approach to improving the transfection properties of cationic lipids has been the synthesis of new kinds of cationic amphipaths. Recently, however, it was found that combining two cationic lipid derivatives having the same head group but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service